Skip to Content
Merck
All Photos(3)

Key Documents

MAK066

Sigma-Aldrich

Lactate Dehydrogenase Activity Assay Kit

sufficient for 500 colorimetric tests

Synonym(s):

LDH Activity Assay Kit, LDH Assay, Lactic Dehydrogenase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 500 colorimetric tests

detection method

colorimetric

relevant disease(s)

hematological disorder; cancer

storage temp.

−20°C

Gene Information

General description

The new Lactate Dehydrogenase Assay Kit, MAK464, is now available! Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate. Cells release LDH into the bloodstream after tissue damage or red blood cell hemolysis. Since LDH is a fairly stable enzyme, it has been widely used to evaluate the presence of damage and toxicity of tissue and cells. LDH is also elevated in certain pathological conditions such as cancer. Quantification of LDH has a broad range of applications.




Application

Lactate Dehydrogenase Activity Assay Kit has been used for measuring the activity of lactate dehydrogenase (LDH) in the cell culture medium.
Lactate Dehydrogenase Activity Assay Kit has been used to determine the concentration of lactate dehydrogenase in samples.

Principle

In this kit, LDH reduces NAD to NADH, which is specifically detected by colorimetric (450 nm) assay. The LDH Activity Assay kit quantifies LDH activity in variety of biological samples. The assay is quick, convenient, and sensitive.

replaced by

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adjimon G Lokossou et al.
Biology of reproduction, 102(1), 185-198 (2019-07-19)
Modulation of the activation status of immune cell populations during pregnancy depends on placental villous cytotrophoblast (VCT) cells and the syncytiotrophoblast (STB). Failure in the establishment of this immunoregulatory function leads to pregnancy complications. Our laboratory has been studying Syncytin-2
Eleonora Cavallari et al.
Frontiers in oncology, 10, 497-497 (2020-05-05)
Nuclear Magnetic Resonance allows the non-invasive detection and quantitation of metabolites to be carried out in cells and tissues. This means that that metabolic changes can be revealed without the need for sample processing and the destruction of the biological
Potassium Bisperoxo(1,10-phenanthroline)oxovanadate (bpV(phen)) Induces Apoptosis and Pyroptosis and Disrupts the P62-HDAC6 Protein Interaction to Suppress the Acetylated Microtubule-dependent Degradation of Autophagosomes.
Chen Q, et al.
The Journal of Biological Chemistry, jbc-M115 (2015)
Francesca Cappellini et al.
Nanomaterials (Basel, Switzerland), 10(4) (2020-04-02)
Relevant in vitro assays that can simulate exposure to nanoparticles (NPs) via inhalation are urgently needed. Presently, the most common method employed is to expose lung cells under submerged conditions, but the cellular responses to NPs under such conditions might
Sayanta Dutta et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 118, 303-316 (2018-05-16)
Arsenic is a potent inducer of several acute and chronic nephrotoxic disorders. It promotes deleterious phenomenon like oxidative stress, inflammation, cell death and altered glucose uptake leading to distorted kidney homeostasis that end up in chronic kidney disease. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service