Skip to Content
Merck
All Photos(2)

Key Documents

11465007001

Roche

Cell Proliferation Kit I (MTT)

Synonym(s):

mtt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

Quality Level

usage

sufficient for ≤2,500 tests

packaging

pkg of 1 kit

manufacturer/tradename

Roche

storage condition

protect from light

technique(s)

tissue culture: suitable

λmax

550-600 nm

application(s)

cell analysis
detection

detection method

colorimetric

storage temp.

−20°C

General description

The Cell Proliferation Kit I (MTT) is a colorimetric assay for the nonradioactive quantification of cellular proliferation, viability, and cytotoxicity. Sample material is either adherent or suspension cells cultured in 96-well microplates.

Colorimetric assays analyze the number of viable cells by the cleavage of tetrazolium salts added to the culture medium. This technique requires neither washing nor harvesting of cells, and the complete assay, from microculture to data analysis by an ELISA reader, is performed in the same microplate.
MTT was the first tetrazolium salt described. It is cleaved to formazan by enzymes of the endoplasmic reticulum. This bioreduction occurs in viable cells only, and is related to NAD(P)H production through glycolysis. Therefore, the amount of formazan dye formed directly correlates to the number of metabolically active cells in the culture.

Colorimetric assay (MTT based) for the nonradioactive quantification of cellular proliferation, viability, and cytotoxicity.

Application

The Cell Proliferation Kit I (MTT) is used for the nonradioactive, spectrophotometric quantification of cell proliferation and viability in cell populations using the 96-well-plate format. It can be used for:
  • Measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients
  • Analysis of cytotoxic and cytostatic compounds, such as anti-cancer drugs and other pharmaceutical compounds
  • Assessment of growth-inhibitory antibodies and physiological mediators
  • Testing of biocompatibility of various scaffolds, employed in bone tissue engineering, for bone cell growth

Features and Benefits

  • Safe and easy: Eliminate radioactive isotopes, washing steps, and additional reagents.
  • Accurate: The absorbance obtained strongly correlates to the cell number.
  • Sensitive: Detect low cell numbers.
  • Fast: Process a large number of samples using a multi-well ELISA reader.
The assay is based on the cleavage of the tetrazolium salt MTT in the presence of an electron-coupling reagent. The water-insoluble formazan salt produced must be solubilized in an additional step. Cells grown in a 96-well tissue culture plate are incubated with the MTT solution for approximately 4 hours. After this incubation period, a water-insoluble formazan dye is formed. After solubilization, the formazan dye is quantitated using a scanning multi-well spectrophotometer (ELISA reader). The measured absorbance directly correlates to the number of viable cells.
Cell proliferation and viability assays are of particular importance for routine applications in cell biology. Tetrazolium salts (e.g., MTT, XTT, WST-1) are particularly useful for this type of analysis. Tetrazolium salts are cleaved to formazan by the succinate-tetrazolium reductase system (EC 1.3.99.1) which belongs to the respiratory chain of the mitochondria, and is only active in metabolically intact cells (see below).

Packaging

1 kit containing 2 components.

Preparation Note

Storage conditions (working solution): 2 to 8 °C
After thawing, the MTT labeling reagents may be stored protected from light at 2 to 8 °C for up to 4 weeks, in which case reagent filtration through a 0.2 μm pore size membrane is recommended.

Kit Components Only

Product No.
Description

  • MTT Labeling Reagent

  • Solubilization Solution

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Tajiri et al.
CNS neuroscience & therapeutics, 22(4), 306-315 (2016-02-05)
Exportin 1 (XPO1/CRM1) plays prominent roles in the regulation of nuclear protein export. Selective inhibitors of nuclear export (SINE) are small orally bioavailable molecules that serve as drug-like inhibitors of XPO1, with potent anti-cancer properties. Traumatic brain injury (TBI) presents
Timothy Douglas et al.
Clinical oral implants research, 21(3), 262-267 (2009-12-05)
Hydroxyapatite (HA) and tricalcium phosphate (TCP) are two very common ceramic materials for bone replacement. A recently developed material for bone replacement is CeraBall, which is a mixed HA-TCP scaffold available as porous spherical scaffolds of diameter 4 and 6
Yuji Kaneko et al.
PloS one, 7(10), e47583-e47583 (2012-10-19)
This study was designed to reveal the therapeutic regimen and mechanism of action underlying hypothermia treatment in combination with stem cell transplantation for ameliorating neonatal hypoxic-ischemic-like injury. Primary rat neurons were exposed to oxygen-glucose deprivation (OGD), which produced hypoxic-ischemic-like injury
Ali Abolhassani et al.
Journal of Cancer, 5(7), 537-547 (2014-07-25)
Fibroblastic growth factor-10 (FGF-10) has an important role in type I epithelial mesenchymal transition (EMT) during the embryonic period of life (gastrulation). Since EMT has a critical role during cancer cells invasion and metastasis (type III) this study sought to
Zhifeng Liu et al.
Molecular medicine reports, 14(1), 1007-1013 (2016-05-26)
The present study aimed to investigate vascular endothelial apoptosis, and the regulatory molecules involved in the condition of heatstroke caused by direct hyperthermia due to high core temperature and gut‑derived endotoxemia. Human umbilical vascular endothelial cells (HUVECs) were isolated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service