Skip to Content
Merck
All Photos(1)

Key Documents

PZ0149

Sigma-Aldrich

CP-868388

≥98% (HPLC)

Synonym(s):

(S)-2-(3-(1-((4-Isopropylbenzyloxy)carbonyl)piperidin-3-yl)phenoxy)-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33NO5
CAS Number:
Molecular Weight:
439.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -50 to -65°, c = 0.2 in methanol

color

white to off-white

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(COC(=O)N2CCC[C@H](C2)c3cccc(OC(C)(C)C(O)=O)c3)cc1

InChI

1S/C26H33NO5/c1-18(2)20-12-10-19(11-13-20)17-31-25(30)27-14-6-8-22(16-27)21-7-5-9-23(15-21)32-26(3,4)24(28)29/h5,7,9-13,15,18,22H,6,8,14,16-17H2,1-4H3,(H,28,29)/t22-/m1/s1

InChI key

CSLFIHDRJSTULR-JOCHJYFZSA-N

Biochem/physiol Actions

CP-868388 is a PPARα agonist.
CP-868388 is a potent PPARα agonist with a Ki of 10.8 nM.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service