Skip to Content
Merck
All Photos(1)

Key Documents

PZ0173

Sigma-Aldrich

CP-775146

≥98% (HPLC)

Synonym(s):

(S)-2-(3-(1-(2-(4-isopropylphenyl)acetyl)piperidin-3-yl)phenoxy)-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33NO4
CAS Number:
Molecular Weight:
423.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -75 to -83° in methanol

color

white to tan

solubility

DMSO: ≥80 mg/mL

storage temp.

−20°C

SMILES string

CC(C)C1=CC=C(CC(N2CCC[C@@H](C3=CC(OC(C)(C)C(O)=O)=CC=C3)C2)=O)C=C1

InChI

1S/C26H33NO4.Na/c1-18(2)20-12-10-19(11-13-20)15-24(28)27-14-6-8-22(17-27)21-7-5-9-23(16-21)31-26(3,4)25(29)30;/h5,7,9-13,16,18,22H,6,8,14-15,17H2,1-4H3,(H,29,30);/q;+1/p-1/t22-;/m1./s1

InChI key

YQAGREGJJZRFDN-VZYDHVRKSA-M

Biochem/physiol Actions

CP-775146 is a potent and selective PPARα agonist.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uxía Nogueira-Recalde et al.
EBioMedicine, 45, 588-605 (2019-07-10)
Ageing-related failure of homeostasis mechanisms contributes to articular cartilage degeneration and osteoarthritis (OA), for which disease-modifying treatments are not available. Our objective was to identify molecules to prevent OA by regulating chondrocyte senescence and autophagy. Human chondrocytes with IL-6 induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service