Skip to Content
Merck
All Photos(3)

Documents

A78403

Sigma-Aldrich

4-Aminopyridine

98%

Synonym(s):

4-Pyridinamine, 4-Pyridylamine, 4AP, Fampridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

General description

4-aminopyridine is a ligand and an ion channel modulator used as an acetylcholine release enhancing agent.

Application

4-Aminopyridine (4-AP) can be used as a:
  • Catalyst in the regioselective acylation of N-tosylhydrazide.
  • Starting material in the synthesis of 4-aminopyridine derivatives for neurological disorder studies.
  • Precursor for the synthesis of enantiomerically pure 4-(pyrrolidino)pyridine (PPY) derivatives by cyclocondensation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of C 2-symmetric analogues of 4-(pyrrolidino) pyridine: new chiral nucleophilic catalysts
Spivey AC, et al.
Journal of the Chemical Society, Part 1, 1(20), 3460-3468 (2000)
Development of novel 4-aminopyridine derivatives as potential treatments for neurological injury and disease
Smith DT, et al.
European Journal of Medicinal Chemistry, 40(9), 908-917 (2005)
Effect of protonated aminopyridines on the structural divergences of M (II)--malonate complexes (M= Cu, Ni, Co)
Amrita D et al.
Polyhedron, 29, 1317-1325 (2010)
4-Aminopyridine catalyzed direct and regioselective acylation of N-tosylhydrazide
Namba K, et al.
Organic Letters, 11(21), 4970-4973 (2009)
4-Aminopyridine derivatives with antiamnesic activity
Aldo A et al.
European Journal of Medicinal Chemistry, 35, 77-82 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service