Skip to Content
Merck
All Photos(2)

Key Documents

PKH67GL

Sigma-Aldrich

PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling

Distributed for Phanos Technologies

Synonym(s):

Green PKH membrane labeling kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

Quality Level

packaging

pkg of 1 kit

storage condition

protect from light

fluorescence

λex 490 nm; λem 502 nm (PKH67 dye)

detection method

fluorometric

shipped in

ambient

storage temp.

room temp

Application

PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling has been used:
  • To label and further investigate the exosomes expelled from cells in triple-negative breast cancer condition.
  • To label apoptotic cells, in order to study the role of ανβ5 receptor in both binding and internalization of apoptotic cells.
  • In fluorescence imaging.

This kit is for general cell membrane labeling. PKH67 has a longer aliphatic carbon tail than PKH1 and PKH2, two other green dyes previously described for in vitro and in vivo cell tracking. Based on the longer tail length, in-house studies have consistently shown reduced cell-cell transfer for PKH67 as compared to PKH2.
Slow loss of fluorescence has been observed in in vivo studies using PKH1 and PKH2. PKH67 may exhibit similar properties since this behavior appears to be characteristic of green cell linker dyes, but not red cell linker dyes. Correlation of in vitro cell membrane retention with in vivo fluorescence half life in non-dividing cells predicts an in vivo fluorescence half life of 10-12 days for PKH67. Other green cell linker dyes with similar half lives have been used to monitor in vivo lymphocyte and macrophage trafficking over periods of 1-2 months, suggesting that PKH67 will also be useful for in vivo tracking studies of moderate length.

Linkage

For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.

Legal Information

CellVue is a registered trademark of Phanos Technologies

Kit Components Only

Product No.
Description

  • Diluent C 6 x 10

  • PKH67 Cell Linker in ethanol .5 mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exosomes from triple-negative breast cancer cells can transfer phenotypic traits representing their cells of origin to secondary cells
O Brien K, et al.
European Journal of Cancer, 49(8), 1845-1859 (2013)
Anjali P Kusumbe et al.
Stem cells (Dayton, Ohio), 27(3), 498-508 (2009-03-04)
Recruitment and localization of endothelial precursors within tumors is a potential area for the development of therapeutics, because their functional contribution to tumor vasculature is realized to be important for cancer cell survival. However, the exact nature of the recruited
Nobuyoshi Kosaka et al.
The Journal of biological chemistry, 287(2), 1397-1405 (2011-11-30)
Normal epithelial cells regulate the secretion of autocrine and paracrine factors that prevent aberrant growth of neighboring cells, leading to healthy development and normal metabolism. One reason for tumor initiation is considered to be a failure of this homeostatic cell
Hybrid microscaffold-based 3D bioprinting of multi-cellular constructs with high compressive strength: A new biofabrication strategy
Yu Jun T, et al.
Scientific Reports, 6, 39140-39140 (2016)
Dandan Ma et al.
Cells, 9(4) (2020-04-01)
Human salivary histatin 1 (Hst1) and Hst2 exhibit a series of cell-activating properties (e.g., promoting adhesion, spreading, migration and metabolic activity of mammalian cells). In contrast, Hst5 shows an anti-fungal property but no cell-activating properties. Previous findings suggest that their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service