Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.32
UNSPSC Code:
12171500
Quality Level
Application
For use as diluent with PKH26-PCL fluorescent cell linker dyes.
Other Notes
For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.
Legal Information
CellVue is a registered trademark of Phanos Technologies
Distributed for Phanos Technologies, Inc.
Kit Components Only
Product No.
Description
- Diluent B 6 vial(s)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PCLDIL-6X10ML | 04061836853617 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service