Skip to Content
Merck
All Photos(1)

Documents

MIDI26

Sigma-Aldrich

PKH26 Red Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling

Distributed for Phanos Technologies

Synonym(s):

pkh membrane dye, red pkh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

packaging

pkg of 1 kit

manufacturer/tradename

Distributed for Phanos Technologies

storage condition

protect from light

technique(s)

flow cytometry: suitable

fluorescence

λex 551 nm; λem 567 nm (PKH26 dye)

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

ambient

storage temp.

room temp

General description

This kit provides enough dye for up to 50 samples of 20 million cells each and enough diluent for up to 30 samples of 20 million cells each when using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye. Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.

Application

This kit is for general cell membrane labeling, including for in vitro cell labeling, in vitro proliferation studies, and long term in vivo cell tracking.

Linkage

For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.

Legal Information

CellVue is a registered trademark of Phanos Technologies

Kit Components Only

Product No.
Description

  • Diluent C 60 mL

  • PKH26 Cell Linker in Ethanol .2 mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingli Han et al.
Cell death & disease, 11(1), 43-43 (2020-01-24)
Trastuzumab is commonly used in the treatment of human epidermal growth factor receptor-2 positive (HER-2+) breast cancer, but its efficacy is often limited by the emergence of chemoresistance. Recent studies indicate that exosomes act as vehicles for exchange of genetic
Chuang Gao et al.
Aging, 11(24), 12147-12164 (2019-12-17)
Exosomes are small (30-150 nm diameter) lipid bilayer-enclosed vesicles found in all bodily fluids. We investigated whether exosomes play a role in chronic subdural hematoma (CSDH). Exosomes were identified and characterized using transmission electron microscopy and NanoSight particle tracking. The
Andres Jerez et al.
Blood, 122(14), 2453-2459 (2013-08-09)
Large granular lymphocyte leukemia (LGL) is often associated with immune cytopenias and can cooccur in the context of aplastic anemia (AA) and myelodysplastic syndromes (MDS). We took advantage of the recent description of signal transducer and activator of transcription 3
Lars T van der Veken et al.
Journal of immunology (Baltimore, Md. : 1950), 182(1), 92-101 (2008-12-26)
Killer Ig-like receptors (KIR) are expressed by human NK cells and T cells. Although Ag-specific cytolytic activity and cytokine production of KIR(+) T cells can be inhibited by KIR ligation, the effect of KIR on proliferation is unclear. KIR(+) T
S Grecchi et al.
European journal of histochemistry : EJH, 58(4), 2449-2449 (2015-01-13)
The endocytotic pathway involves a complex, dynamic and interacting system of intracellular compartments. PKH26 is a fluorescent dye specific for long-lasting cell membrane labelling which has been successfully used for investigating cell internalization processes, at either flow cytometry or fluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service