Skip to Content
Merck
All Photos(5)

Key Documents

P9404

Sigma-Aldrich

Poly-L-lysine hydrochloride

suitable for cell culture, Mol wt >30,000

Synonym(s):

L-Lysine hydrochloride homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.26

Product Name

Poly-L-lysine hydrochloride, mol wt >30,000

form

powder

Quality Level

mol wt

>30,000

technique(s)

FTIR: suitable
cell culture | mammalian: suitable

color

white to light yellow

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly-lysine with mol. wt. >70,000 is useful in promoting cell adhesion to solid substrates.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dendritic cell-targeted protein vaccines: a novel approach to induce T cell immunity.
Trumpfheller C, Longhi MP, et al.
J. Int. Med., 271(2), 183-193 (2011)
Jérôme Roy
Brain research, 1689, 21-29 (2018-03-27)
Microglia are the main resident immunological cells of the central nervous system (CNS) that are functionally equivalent to macrophages. However, due to the cellular heterogeneity of the brain, it is technically challenging to obtain highly specific, healthy microglia with the
Yen-Wei Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 33(26), 6521-6527 (2017-06-13)
Nanodiamond (ND) is a carbon-based nanomaterial with potential for a wide range of biological applications. One of such applications is to facilitate the nucleation of protein crystals in aqueous solution. Here, we show that NDs (nominal diameters of 30 and
C M Gardner et al.
Journal of materials science. Materials in medicine, 23(1), 181-193 (2011-12-20)
Calcium alginate/poly-L-lysine beads were coated with either 50% hydrolyzed poly(methyl vinyl ether-alt-maleic anhydride) (PMM(50)), or with poly(vinyl dimethyl azlactone-co-methacrylic acid) (50:50, PMV(50)), to form covalently shell-crosslinked capsules, and compared with analogous capsules coated with sodium alginate. All capsule types were
Arlin Jose Amali et al.
Chemical communications (Cambridge, England), 48(6), 856-858 (2011-11-30)
Counter-ion condensation of Poly(L-Lysine) in the presence of pyranine-3 generates spherical coacervates, which then template the assembly of silica nanoparticles to form microcapsule structures that dynamically control the optical ratiometric sensing of both the change in pH and release of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service