Skip to Content
Merck
All Photos(3)

Key Documents

P7762

Sigma-Aldrich

Poly-L-arginine hydrochloride

mol wt 15,000-70,000

Synonym(s):

L-Arginine homopolymer hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

form

powder

mol wt

15,000-70,000

color

white to off-white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

Cl.NC(CCCNC(N)=N)C(O)=O

InChI

1S/C18H38N12O4.ClH/c19-10(4-1-7-26-16(20)21)13(31)29-11(5-2-8-27-17(22)23)14(32)30-12(15(33)34)6-3-9-28-18(24)25;/h10-12H,1-9,19H2,(H,29,31)(H,30,32)(H,33,34)(H4,20,21,26)(H4,22,23,27)(H4,24,25,28);1H/t10-,11-,12-;/m0./s1

InChI key

PQEISBBDIRAKGR-LFELFHSZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly-L-arginine hydrochloride (PLA) is a homopolyamino acid. PLA enhances membrane permeability compared to poly-L-lysine. It has applications as drug carrier, neuroprotective agent and in anticancer vaccines preparations. Peptide vaccines in conjugation with PLA promotes antigen-presenting cells (APCs) to generate T cell and elicit immunostimulant functionality.

Application

Poly-L-arginine hydrochloride has been used:
  • to generate complex with stimulatory RNA isRNA9.2 as in neutrophils
  • used in ribosomal RNA transfection into human monocytic THP-1 cells
  • for coating alginate capsule beads

Quality

Contains 3% or less ornithine residue.

Analysis Note

Molecular weight based on viscosity of precursor poly-L-ornithine. Also assayed by MALLS.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors influencing the mechanical stability of alginate beads applicable for immunoisolation of mammalian cells
Bhujbal SV, et al.
Journal of the Mechanical Behavior of Biomedical Materials, 37, 196-208 (2014)
Vaccination with poly-L-arginine as immunostimulant for peptide vaccines: induction of potent and long-lasting T-cell responses against cancer antigens
Mattner F, et al.
Cancer Research, 62(5), 1477-1480 (2002)
RNA of Enterococcus faecalis strain EC-12 is a major component inducing interleukin-12 production from human monocytic cells
Nishibayashi R, et al.
PLoS ONE, 10(6), e0129806-e0129806 (2015)
Antibacterial activity of poly-l-arginine under different conditions
Sepahi M , et al.
Iranian Journal of Microbiology, 9(2), 103-103 (2017)
Kotaro Sakamoto et al.
Frontiers in pharmacology, 12, 751587-751587 (2021-11-26)
Worldwide, more than 20 million people suffer from schizophrenia, but effective and definitive new therapeutic drugs/treatments have not been established. Vasoactive intestinal peptide receptor 2 (VIPR2) might be an attractive drug target for the treatment of schizophrenia because both preclinical and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service