Skip to Content
Merck
All Photos(3)

Key Documents

P4663

Sigma-Aldrich

Poly-L-arginine hydrochloride

mol wt 5,000-15,000

Synonym(s):

L-Arginine homopolymer hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

form

powder

mol wt

5,000-15,000

color

white to off-white

application(s)

cell analysis
detection

storage temp.

−20°C

SMILES string

Cl.NC(CCCNC(N)=N)C(O)=O

InChI

1S/C18H38N12O4.ClH/c19-10(4-1-7-26-16(20)21)13(31)29-11(5-2-8-27-17(22)23)14(32)30-12(15(33)34)6-3-9-28-18(24)25;/h10-12H,1-9,19H2,(H,29,31)(H,30,32)(H,33,34)(H4,20,21,26)(H4,22,23,27)(H4,24,25,28);1H/t10-,11-,12-;/m0./s1

InChI key

PQEISBBDIRAKGR-LFELFHSZSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Interfacial Electrostatics of Poly(vinylamine hydrochloride), Poly(diallyldimethylammonium chloride), Poly-L-lysine, and Poly-L-arginine Interacting with Lipid Bilayers.: This research investigates the interactions between poly-L-arginine hydrochloride and lipid bilayers, providing insights into its potential applications in drug delivery systems (McGeachy et al., 2018). (McGeachy et al., 2018).

Quality

Contains 3% or less ornithine residue.

Analysis Note

Molecular weight based on viscosity of precursor poly-L-ornithine.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramsha Aamir et al.
International journal of molecular sciences, 22(21) (2021-11-14)
Ischemic stroke is the most common type of stroke and thrombolytic therapy is the only approved treatment. However, current thrombolytic therapy with tissue plasminogen activator (tPA) is often hampered by the increased risk of hemorrhage. Plasmin, a direct fibrinolytic, has
Analysis of transient and reversible effects of poly-L-arginine on the in vivo nasal absorption of FITC-dextran in rats.
Ohtake K, Natsume H, Ueda H, et al.
J. Controlled Release, 82, 263-275 (2002)
M Miyamoto et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 52(1), 21-30 (2001-07-05)
The effects of the concentration and molecular weight of poly-L-arginine (poly-L-Arg) on the in vivo nasal absorption of fluorescein isothiocyanate-labeled dextran (MW, 4 kDa, FD-4) in rats were studied. When poly-L-Arg with a range of different molecular weights (MW, 8.9
Keerthi Nadimidla et al.
Biopolymers, 107(9) (2017-05-01)
Tau protein plays a major role in Alzheimer's disease. The tau protein loses its functionality by self-aggregation due to the two six-amino acid sequences VQIVYK and VQIINK of the protein. Hence it is imperative to find therapeutics that could inhibit
Poly-L-arginine enhances paracellular permeability via serine/threonine phosphorylation of ZO-1 and tyrosine dephosphorylation of occludin in rabbit nasal epithelium.
Ohtake K, Maeno T, Ueda H, et al.
Pharmaceut. Res., 20, 1838-1845 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service