Skip to Content
Merck
All Photos(1)

Key Documents

P8765

Sigma-Aldrich

Ammonium pyrrolidinedithiocarbamate

~99%

Synonym(s):

1-Pyrrolidinecarbodithioic acid ammonium salt, APDC, Ammonium pyrrolidinecarbodithioate, PDC, PDTC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9NS2 · NH3
CAS Number:
Molecular Weight:
164.29
Beilstein:
3730472
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77

biological source

microbial

Quality Level

Assay

~99%

form

(powderor powder with lumps)

mp

153-155 °C (lit.)

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

N.SC(=S)N1CCCC1

InChI

1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3

InChI key

MDDIUTVUBYEEEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium pyrrolidinedithiocarbamate is a nuclear factor κB (NF-κB) inhibitor. It functions as a chelating agent, which allows the preconcentration of trace metals from solution. Pyrrolidinedithiocarbamate (PDTC) has antioxidant properties. It inhibits apoptosis in lymphocytes, neurons and vascular endothelial cells.

Application

Ammonium pyrrolidinedithiocarbamate has been used:
  • to inhibit nuclear factor κB (NF-κB) mobilization and TNF (tumor necrosis factor) production in human monocytes
  • to reduce neutrophil activity in rats
  • to prevent Ca2+ influx in adriamycin (ADR)-treated podocytes

to abrogate the microRNA-146a promoter luciferase reporter activity in human brain cells

Biochem/physiol Actions

Prevents induction of nitric oxide synthetase by inhibiting translation of NOS mRNA; induces apoptosis in rat smooth muscle cells and inhibits apoptosis in leukemia HL-60 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An NF-kappaB-sensitive micro RNA-146a-mediated inflammatory circuit in Alzheimer disease and in stressed human brain cells
Lukiw WJ, et al.
The Journal of Biological Chemistry, 283(46), 31315-31322 (2008)
Quan Zhou et al.
Oncogene, 39(7), 1414-1428 (2019-10-30)
Gastric cancer (GC) is characterized by extensive local invasion, distant metastasis and poor prognosis. In most cases, GC progression is associated with aberrant expression of cytokines or activation of signaling cascades mediated by tumor-stroma interactions. However, the mechanisms by which
Induction of apoptosis by pyrrolidinedithiocarbamate and N-acetylcysteine in vascular smooth muscle cells
Tsai JC, et al.
The Journal of Biological Chemistry, 271(7), 3667-3670 (1996)
Electron transport chain inhibitors induce microglia activation through enhancing mitochondrial reactive oxygen species production
Ye J, et al.
Experimental Cell Research, 340(2), 315-326 (2016)
Multi-element preconcentration of heavy metal ions from aqueous solution by APDC impregnated activated carbon
Daorattanachai P, et al.
Talanta, 67(1), 59-64 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service