Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium pyrrolidinedithiocarbamate
Synonym(s):
1-Pyrrolidinecarbodithioic acid ammonium salt, APDC, Ammonium pyrrolidinecarbodithioate, PDC, PDTC
Linear Formula:
C
5
H
9
NS
2
· NH
3
CAS Number:
5108-96-3
Molecular Weight:
164.29
EC Number:
225-834-4
Products
Brand
Sigma-Aldrich (2)
Biological Source
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P8765
Ammonium pyrrolidinedithiocarbamate,
~99%
Expand
View Pricing
09935
Ammonium pyrrolidinedithiocarbamate,
purum p.a., ≥98.0% (NT)
Expand
View Pricing