Skip to Content
Merck
All Photos(1)

Key Documents

W273104

Sigma-Aldrich

4-Methyl-2-pentanone

≥99%, FCC

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
FEMA Number:
2731
Beilstein:
605399
EC Number:
Council of Europe no.:
151
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.017
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

Assay

≥99%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; fruity; spicy

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-2-pentanone has been identified in the volatile fraction of lemon essence, Italian chestnut and crabmeat.

Application


  • Thermodynamic Analysis of Eplerenone in 13 Pure Solvents at Temperatures from 283.15 to 323.15 K.: This study provides a detailed analysis of the solubility behavior and thermodynamics of Eplerenone, offering valuable insights for its application in solvent selection and pharmaceutical formulation (Liu et al., 2024).

  • Development and validation of a high-throughput headspace solid-phase microextraction gas chromatography-mass spectrometry methodology for target and suspect determination of honey volatiles.: This article presents a method for the analysis of volatile compounds in honey, where 4-Methyl-2-pentanone is used to enhance the detection of aroma compounds critical for quality control and authenticity assessments (Gialouris et al., 2023).
  • .

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compound analysis of SPME headspace and extract samples from roasted Italian chestnuts (Castanea sativa Mill.) using GC-MS.
Krist S, et al.
Food Sci. Technol., 219(5), 470-473 (2004)
Analysis of crabmeat volatile compounds.
Matiella JE & Hsieh TCY.
Journal of Food Science, 55(4), 962-966 (1990)
Analysis of flavor constituents from lemon and lime essence.
Moshonas MG and Shaw PE.
Journal of Agricultural and Food Chemistry, 20(5), 1029-1030 (1972)
H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Mao Ting et al.
The Science of the total environment, 390(1), 97-108 (2007-11-03)
Volatile organic compounds (VOCs) in the air of Beijing City were measured at the heights of 8, 32, 140 and 280 m on the Beijing 325 m meteorological tower in autumn 2005. Concentrations of fifty-five compounds were determined by quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service