Skip to Content
Merck
All Photos(2)

Key Documents

319775

Sigma-Aldrich

Octanophenone

≥99%

Synonym(s):

Caprylophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COC6H5
CAS Number:
Molecular Weight:
204.31
Beilstein:
1868534
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.5044 (lit.)

bp

285-290 °C (lit.)

mp

21.5-23.5 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCCCCCCC(=O)c1ccccc1

InChI

1S/C14H20O/c1-2-3-4-5-9-12-14(15)13-10-7-6-8-11-13/h6-8,10-11H,2-5,9,12H2,1H3

InChI key

UDEVCZRUNOLVLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunta Futagami et al.
Journal of chromatography. A, 1595, 58-65 (2019-02-26)
We report on the ability to change the layer properties of porous layered radially elongated pillar (PLREP) array columns and its relevance to the separation efficiency. The adjustment of the preparation condition resulted in the formation of a 1.2-fold thicker
Gert Desmet et al.
Journal of chromatography. A, 1626, 461339-461339 (2020-08-17)
We report on a systematic and comprehensive (0.7 ≤ k'' ≤ 122) experimental study of the effect of the zone retention factor k'' on eddy dispersion (heddy) in packed bed columns for liquid chromatography. The values for heddy are obtained by subtracting
Sam Wouters et al.
Journal of chromatography. A, 1523, 224-233 (2017-06-18)
The present paper discusses practical aspects of prototyping of microfluidic chips using cyclic olefin copolymer as substrate and the application in high-performance liquid chromatography. The developed chips feature a 60mm long straight separation channel with circular cross section (500μm i.d.)
Shunta Futagami et al.
The Analyst, 144(5), 1809-1817 (2019-01-24)
The performance of a porous-layered radially elongated pillar (PLREP) array column in a commercial nano-LC system was examined by performing separation of alkylphenones and peptides. The mesoporous silica layer was prepared by sol-gel processing of a mixture of tetramethoxysilane and
Gert Desmet et al.
Journal of chromatography. A, 1626, 461279-461279 (2020-08-17)
We report on a comparative study of the basic separation kinetics of commercial packed bed columns and a micro-pillar array column (μPAC) working in the 1-10μL/min flow rate range, i.e., operating in the area of capillary flow LC. This is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service