Skip to Content
Merck
All Photos(1)

Key Documents

C4382

Sigma-Aldrich

Carbamoylcholine chloride

≥98% (titration), crystalline

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride carbamate, Carbachol, Carbamylcholine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOCH2CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
182.65
Beilstein:
3917459
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (titration)

form

crystalline

color

white

mp

210 °C (dec.) (lit.)

solubility

H2O: 1 g/mL
ethanol: 20 mg/mL

SMILES string

[Cl-].C[N+](C)(C)CCOC(N)=O

InChI

1S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1H

InChI key

AIXAANGOTKPUOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Carbamoylcholine chloride has been used:
  • to stimulate insulin secretion in transgenic mice
  • to induce contraction of undifferentiated and differentiated human adipose-derived stem cells (hASCs)
  • to mediate constriction of iris in pupillary light reflex studies in mice

Biochem/physiol Actions

Carbachol elicits tear fluid secretion by interacting with muscarinic receptors. It is an analog of acetylcholine and an antiglaucoma drug. In ciliary processes, carbachol along with nitric oxide (NO) regulates Na+ and K+-ATPase activity. It stimulates insulin vesicles transport to the secretory site to increase insulin secretion.
Non-selective cholinergic agonist that is resistant to the action of cholinesterases; inhibits apoptotic death of cultured neurons, and may induce apoptosis in thymocytes.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic profiling of tissue-engineered blood vessel walls constructed by adipose-derived stem cells
Wang C, et al.
Tissue Engineering: Part A, 19(3-4), 415-425 (2012)
Role of the non-neuronal cholinergic system in the eye: a review
Duncan G and Collison DJ
Life Sciences, 72(18-19), 2013-2019 (2003)
Carbachol and nitric oxide inhibition of Na, K-ATPase activity in bovine ciliary processes
Ellis DZ, et al.
Investigative Ophthalmology & Visual Science, 42(11), 2625-2631 (2001)
Anne Cathrine Nøhr et al.
Scientific reports, 9(1), 3802-3802 (2019-03-09)
GPR139 is an orphan G protein-coupled receptor (GPCR) that is primarily expressed in the brain in regions known to regulate motor control and metabolism. Here, we screened a diverse 4,000 compound library in order to identify GPR139 agonists. We identified
Targeted mutation of the calbindin D28k gene selectively alters nonvisual photosensitivity
Butler MP, et al.
The European Journal of Neuroscience, 33(12), 2299-2307 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service