Skip to Content
Merck
All Photos(1)

Documents

P7173

Sigma-Aldrich

Pyruvate Carboxylase from bovine liver

buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)

Synonym(s):

Pyruvate:CO2 ligase (ADP-forming)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

form

buffered aqueous glycerol solution

specific activity

5-25 units/mg protein (BCA)

concentration

≥0.5 mg/mL

foreign activity

lactic dehydrogenase ≤0.5%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pyruvate is critical for gluconeogenesis, lipogenesis, glyceroneogenesis, neurotransmitter biosynthesis and glucose-induced insulin, and is used to study these processes.
The enzyme from Sigma has been used as a positive control during the assay of pyruvate carboxylase activity in cell-free extracts of Corynebacterium glutamicum.

Biochem/physiol Actions

Pyruvate carboxylase catalyzes the carboxylation of pyruvate to oxaloacetate. Pyruvate carboxylase is a mitochondrial protein that has a biotin prosthetic group that requiries magnesium or manganese and acetyl CoA.

Unit Definition

One unit will convert 1.0 μmole of pyruvate and CO2 to oxalacetate per min at pH 7.8 at 30 °C.

Physical form

Solution in 50% glycerol containing 0.05 M Tris-HCl, pH 7.4, 2 mM magnesium acetate and 1 mM EDTA.

Preparation Note

Affinity purified

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunsook S Jin et al.
Metabolism: clinical and experimental, 62(1), 152-162 (2012-09-18)
A three-day high-fat diet induces hepatic steatosis and hepatic insulin resistance in rats without altering fasting plasma glucose concentration or the rate of glucose production. However, as the nutrient profile available to the liver is substantially altered by a high-fat
Philip Lee et al.
Hepatology (Baltimore, Md.), 57(2), 515-524 (2012-08-23)
The pathogenesis of type 2 diabetes is characterized by impaired insulin action and increased hepatic glucose production (HGP). Despite the importance of hepatic metabolic aberrations in diabetes development, there is currently no molecular probe that allows measurement of hepatic gluconeogenic
M L Lazo de la Vega-Monroy et al.
The Journal of nutritional biochemistry, 24(1), 169-177 (2012-07-31)
Besides its role as a carboxylase cofactor, biotin has a wide repertoire of effects on gene expression, development and metabolism. Pharmacological concentrations of biotin enhance insulin secretion and the expression of genes and signaling pathways that favor islet function in
H M White et al.
Journal of dairy science, 95(3), 1249-1256 (2012-03-01)
Expression of mRNA for pyruvate carboxylase (PC) is elevated at calving and during other physiological states when plasma NEFA concentrations are increased. The objective of this study was to determine the direct effects of fatty acids on expression of PC
Scott B Crown et al.
BMC systems biology, 6, 43-43 (2012-05-18)
¹³C-Metabolic flux analysis (¹³C-MFA) is a standard technique to probe cellular metabolism and elucidate in vivo metabolic fluxes. 13C-Tracer selection is an important step in conducting ¹³C-MFA, however, current methods are restricted to trial-and-error approaches, which commonly focus on an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service