Skip to Content
Merck
All Photos(1)

Documents

10109045001

Roche

Pyruvate Kinase (PK)

from rabbit muscle

Synonym(s):

PK

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

rabbit muscle

Quality Level

form

solution

specific activity

~200 units/mg protein (At 25 °C with PEP as the substrate.)

packaging

pkg of 1 mL (10 mg)

manufacturer/tradename

Roche

optimum pH

7.0-7.5

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Related Categories

General description

Pyruvate kinase has a molar mass of 237,000 and exists as a tetramer. Each polypeptide chain of this tetramer has a molar mass of 57,200. The enzyme contains two identical catalytic particles called protomers. Each of these protomers contains two polypeptide chains. Each protomer contains one site each for Mn2+ and phosphoenolpyruvate.

Specificity

Specific activity: Approximately 200 U/mg at +25°C with PEP as the substrate.

Application

Pyruvate Kinase (PK) has been used as a sample in ATPase Assays.
Pyruvate Kinase (PK) has been used in the kinase assay.

Biochem/physiol Actions

Pyruvate kinase catalyzes the irreversible conversion of P-enolpyruvate and ADP to pyruvate and ATP with the utilization of a proton. The first step is the transfer of phosphate group from P-enolpyruvate to ADP with the formation of bound enolate of pyruvate and ATP. In the second step, a proton is added to enolate to generate the keto form of pyruvate. The enzyme also exhibits other activities, such as ATP- and bicarbonate-dependent ATPase, phosphorylation of fluoride and hydroxylamine, ATP-dependent phosphorylation of glycolate, and decarboxylation of oxaloacetate.

Quality

Contaminants: <0.001% GK, <0.002% HK, “NADH oxidase”, and ATPase, each, <0.01% enolase, LDH, and myokinase, each.

Physical form

Solution in 50% glycerol (v/v), pH approximately 6

Preparation Note

Activator: PK requires Mg2+ (or Mn2+, Co2+) and K+ (or NH4+, Rb+) for full activity.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo evidence for ATPase-dependent DNA translocation by the Bacillus subtilis SMC condensin complex
Wang X, et al.
Molecular Cell, 71(5), 841-847 (2018)
Matthias M Schneider et al.
Nature communications, 12(1), 5999-5999 (2021-10-16)
Molecular chaperones contribute to the maintenance of cellular protein homoeostasis through assisting de novo protein folding and preventing amyloid formation. Chaperones of the Hsp70 family can further disaggregate otherwise irreversible aggregate species such as α-synuclein fibrils, which accumulate in Parkinson's
The structure of the R2TP complex defines a platform for recruiting diverse client proteins to the HSP90 molecular chaperone system
Rivera-Calzada A, et al.
Structure, 25(7), 1145-1152 (2017)
Ryo R Watanabe et al.
iScience, 26(5), 106626-106626 (2023-05-17)
F1-ATPase (F1) is an ATP-driven rotary motor protein ubiquitously found in many species as the catalytic portion of FoF1-ATP synthase. Despite the highly conserved amino acid sequence of the catalytic core subunits: α and β, F1 shows diversity in the
Metabolic control and structure of glycolytic enzymes. 3. Dissociation and subunit structure of rabbit muscle pyruvate kinase.
M A Steinmetz et al.
Biochemistry, 5(4), 1399-1405 (1966-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service