Skip to Content
Merck
All Photos(2)

Key Documents

P6636

Sigma-Aldrich

L-α-Phosphatidylinositol from Glycine max (soybean)

~50% (TLC)

Synonym(s):

1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol), Afamelanotide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
8535268
EC Number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

soybean

form

crude powder

concentration

~50% (TLC)

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@@H]1[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C45H87O13P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(46)55-35-37(36-56-59(53,54)58-45-43(51)41(49)40(48)42(50)44(45)52)57-39(47)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40-45,48-52H,3-36H2,1-2H3,(H,53,54)/t37-,40?,41-,42+,43-,44-,45?/m1/s1

InChI key

FQZQXPXKJFOAGE-KAEDGTSCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Phospholipid membrane component, precursor of inositol phosphates.

Quality

May contain phosphatidic acid and phosphatidyl ethanolamine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adi Haber et al.
PLoS pathogens, 13(1), e1006161-e1006161 (2017-01-24)
The high environmental adaptability of bacteria is contingent upon their ability to sense changes in their surroundings. Bacterial pathogen entry into host poses an abrupt and dramatic environmental change, during which successful pathogens gauge multiple parameters that signal host localization.
T Chakraborty et al.
Journal of bacteriology, 174(2), 568-574 (1992-01-01)
The prfA gene of Listeria monocytogenes encodes a protein that activates transcription of the listeriolysin gene (lisA). In order to explore the role of the prfA gene product in the pathogenesis of listerial infection, we constructed a site-directed insertion mutation
Tamar Burg-Golani et al.
Journal of bacteriology, 195(23), 5262-5272 (2013-09-24)
Listeria monocytogenes is a Gram-positive human intracellular pathogen that infects diverse mammalian cells. Upon invasion, L. monocytogenes secretes multiple virulence factors that target host cellular processes and promote infection. It has been presumed, but was not empirically established, that the
Qiangjun Zhou et al.
Nature communications, 5, 3552-3552 (2014-03-29)
Phosphatidylinositol 4-kinase IIα (PI4KIIα), a membrane-associated PI kinase, plays a central role in cell signalling and trafficking. Its kinase activity critically depends on palmitoylation of its cysteine-rich motif (-CCPCC-) and is modulated by the membrane environment. Lack of atomic structure
Lior Lobel et al.
Molecular microbiology, 95(4), 624-644 (2014-11-29)
Metabolic adaptations are critical to the ability of bacterial pathogens to grow within host cells and are normally preceded by sensing of host-specific metabolic signals, which in turn can influence the pathogen's virulence state. Previously, we reported that the intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service