Skip to Content
Merck
All Photos(2)

Key Documents

P9511

Sigma-Aldrich

3-sn-Phosphatidic acid sodium salt from egg yolk lecithin

≥98%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphate sodium salt, L-α-Phosphatidic acid sodium salt, PA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].OC[C@@H](O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na/c4-1-3(5)2-9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;/q;2*+1/p-2/t3-;;/m1../s1

InChI key

GEKBIENFFVFKRG-HWYNEVGZSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Phosphatidic acid is obtained from phosphatidylcholine with the help of phospholipase D.

Application

3-sn-Phosphatidic acid sodium salt from egg yolk lecithin
  • in the reaction mixture for CDS (cytidine triphosphate:phosphatidate cytidyltransferase) assay and reference lipid for thin layer chromatography (TLC)
  • as a standard to compare the generated radiolabeled phosphatidic acid (PA)
  • to examine its effects on At-Rac2–associated kinase activity

Biochem/physiol Actions

Phosphatidic acid might serve as a modulator of phagocyte respiratory burst. It also acts as a precursor of diacylglycerol, phosphatidic acid and lysophosphatidic acid.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases, Phosphoinositide Kinases and Phosphoprotein Phosphatases (Serine/Threonine) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Prepared from L-α-phosphatidylcholine by hydrolysis with cabbage phospholipase D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rac homologues and compartmentalized phosphatidylinositol 4, 5-bisphosphate act in a common pathway to regulate polar pollen tube growth
Kost B, et al.
The Journal of cell biology, 145(2), 317-330 (1999)
Complementary DNAs encoding eukaryotic-type cytidine-5 [prime]-diphosphate-diacylglycerol synthases of two plant species
Kopka J, et al.
Plant Physiology, 113(3), 997-1002 (1997)
Daniel Pastor-Flores et al.
Biochimica et biophysica acta, 1861(3), 249-259 (2016-01-09)
Pkh is the yeast ortholog of the mammalian 3-phosphoinositide-dependent protein kinase 1 (PDK1). Pkh phosphorylates the activation loop of Ypks, Tpks, Sch9 and also phosphorylates the eisosome components Lsp1 and Pil1, which play fundamental roles upstream of diverse signaling pathways
Stefano Mancuso et al.
Plant signaling & behavior, 2(4), 240-244 (2007-07-01)
Auxin (IAA) is versatile signalling molecule of plants, currently classified as plant hormone. But there are data suggesting that auxin is acting also as plant-specific morphogen, electric-responses inducing transmitter, and as general signalling molecule used for plant-bacteria communication. Our previous
Glycerolipids in signal transduction
Lambeth JD, et al.
New Biotechnology, 31, 237-255 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service