P2517
L-α-Phosphatidylinositol ammonium salt solution from bovine liver
10 mg/mL in chloroform, ≥98% (TLC)
Synonym(s):
1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol), PI, PtdIns
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Related Categories
Biochem/physiol Actions
Phosphatidylinositol is a phosphoglyceride, containing inositol as a cyclohexanol head group. The phosphatidylinositol signaling pathway is also called as double signaling pathway as it generates two secondary messengers. It is associated with a number of physiological processes in plants. This includes the development of pollen and vascular organs, root and root hair growth, hormone response and stress tolerance.
Phospholipid membrane component, precursor of inositol phosphates.
Features and Benefits
This compound is featured on the Phosphoinositide Kinases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Other Notes
Contains primarily stearic and arachidonic acids.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cell Biology, 471-471 (2007)
Functional Characterization of Arabidopsis Phosphatidylinositol Monophosphate 5-kinase 2 in Lateral Root Development, Gravitropism and Salt Tolerance, 2-2 (2014)
Sarah J Tilley et al.
Structure (London, England : 1993), 12(2), 317-326 (2004-02-14)
Phosphatidylinositol transfer protein alpha (PITPalpha) selectively transports and promotes exchange of phosphatidylinositol (PI) and phosphatidylcholine (PC) between lipid bilayers. In higher eukaryotes PITPalpha is required for cellular functions such as phospholipase C-mediated signaling, regulated exocytosis, and secretory vesicle formation. We
S J Heydrick et al.
The Journal of clinical investigation, 91(4), 1358-1366 (1993-04-01)
Activation of phosphatidylinositol-3-kinase (PI3K) is one of the earliest postreceptor events in the insulin signaling pathway. Incubation of soleus muscles from lean mice with 50 nM insulin caused a 3-10-fold increase in antiphosphotyrosine-immunoprecipitable PI3K (antiPTyr-PI3K) activity within 2 min in
D Voelker et al.
Biophysical journal, 70(2), 818-830 (1996-02-01)
We have measured the distribution of the hexavalent ruthenium red cation (RuR) between water and phospholipid membranes, have shown the critical importance of membrane negative surface charge for RuR binding, and determined the association constant of RuR for different phospholipid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service