Skip to Content
Merck
All Photos(1)

Key Documents

P6386

Sigma-Aldrich

L-α-Phosphatidylethanolamine from egg yolk

Type III, 10 mg/mL in chloroform, ≥97%, solution

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

biological source

egg yolk

Quality Level

type

Type III

Assay

≥97%

form

solution

concentration

10 mg/mL in chloroform

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCC)(OCCN)O

InChI

1S/C40H80NO8P/c1-3-5-7-9-11-13-15-17-18-19-20-21-23-25-27-29-31-33-40(43)49-38(37-48-50(44,45)47-35-34-41)36-46-39(42)32-30-28-26-24-22-16-14-12-10-8-6-4-2/h38H,3-37,41H2,1-2H3,(H,44,45)/t38-/m1/s1

InChI key

NJGIRBISCGPRPF-KXQOOQHDSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Purity when prepared is 98-99%. Solutions exposed to room temperature can decompose 0.3-0.5% per day.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ram Dvash et al.
Journal of controlled release : official journal of the Controlled Release Society, 170(2), 295-305 (2013-06-15)
In spite of significant insolubility and toxicity, carbon nanotubes (CNTs) erupt into the biomedical research, and create an increasing interest in the field of nanomedicine. Single-walled CNTs (SWCNTs) are highly hydrophobic and have been shown to be toxic while systemically
Allyson A West et al.
The American journal of clinical nutrition, 97(4), 718-727 (2013-03-01)
Phosphatidylcholine (PC) produced via the S-adenosylmethionine-dependent phosphatidylethanolamine (PE) N-methyltransferase (PEMT) pathway is enriched with docosahexaenoic acid (DHA). DHA plays a critical role in fetal development and is linked to health endpoints in adulthood. It is unknown whether choline, which can
Faraz Quazi et al.
The Journal of biological chemistry, 288(48), 34414-34426 (2013-10-08)
ABCA1, ABCA7, and ABCA4 are members of the ABCA subfamily of ATP-binding cassette transporters that share extensive sequence and structural similarity. Mutations in ABCA1 cause Tangier disease characterized by defective cholesterol homeostasis and high density lipoprotein (HDL) deficiency. Mutations in
Christin Stegemann et al.
Circulation, 129(18), 1821-1831 (2014-03-14)
The bulk of cardiovascular disease risk is not explained by traditional risk factors. Recent advances in mass spectrometry allow the identification and quantification of hundreds of lipid species. Molecular lipid profiling by mass spectrometry may improve cardiovascular risk prediction. Lipids
Heike Anders et al.
International journal of systematic and evolutionary microbiology, 64(Pt 4), 1264-1270 (2014-01-16)
A strictly aerobic, thermophilic, moderately acidophilic, non-spore-forming bacterium, strain P373(T), was isolated from geothermally heated soil at Waikite, New Zealand. Cells were filamentous rods, 0.2-0.4 µm in diameter and grew in chains up to 80 µm in length. On the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service