Skip to Content
Merck
All Photos(1)

Key Documents

P2621

Sigma-Aldrich

Phosphomannose Isomerase from Escherichia coli

recombinant, expressed in E. coli, ammonium sulfate suspension, ≥50 units/mg protein

Synonym(s):

D-Mannose-6-phosphate ketol-isomerase, Mannose Phosphate Isomerase, PMI

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

ammonium sulfate suspension

specific activity

≥50 units/mg protein

storage temp.

2-8°C

Related Categories

Application

PMI is used to study cell wall synthesis and energy production. PMI has been used to study how EDTA and metal ions, such as Zn++, Co++, Fe++, Mn++ and Cu++., can affect recovery and thermal stability. It may be used to study PMI′s effect on various alginate biosynthetic enzymes such as phosphomannomutase (PMM), GDP-mannose pyrophosphorylase (GMP), and GDP-mannose dehydrogenase (GMD).

Biochem/physiol Actions

Phosphomannose Isomerase (PMI) catalyses the interconversion of mannose 6-phosphate (Man-6-P) and fructose 6-phosphate (Fru-6-P), which provides a link between glucose metabolism and mannosylation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

One unit will convert 1.0 μmole of D-mannose 6-phosphate to D-fructose 6-phosphate per min at pH 7.6 at 25 °C, using a coupled enzyme system with phosphoglucose isomerase and glucose-6-phosphate dehydrogenase.

Physical form

Supplied as a suspension in 3.2 M ammonium sulfate

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell Dahl et al.
Journal of medicinal chemistry, 54(10), 3661-3668 (2011-05-05)
We report the discovery and validation of a series of benzoisothiazolones as potent inhibitors of phosphomannose isomerase (PMI), an enzyme that converts mannose-6-phosphate (Man-6-P) into fructose-6-phosphate (Fru-6-P) and, more importantly, competes with phosphomannomutase 2 (PMM2) for Man-6-P, diverting this substrate
Craig T Narasaki et al.
PloS one, 6(10), e25514-e25514 (2011-11-09)
Coxiella burnetii, the etiologic agent of human Q fever, is a gram-negative and naturally obligate intracellular bacterium. The O-specific polysaccharide chain (O-PS) of the lipopolysaccharide (LPS) of C. burnetii is considered a heteropolymer of the two unusual sugars β-D-virenose and
Stéphanie Desvergnes et al.
Bioorganic & medicinal chemistry, 20(4), 1511-1520 (2012-01-25)
In the design of inhibitors of phosphosugar metabolizing enzymes and receptors with therapeutic interest, malonate has been reported in a number of cases as a good and hydrolytically-stable surrogate of the phosphate group, since both functions are dianionic at physiological
Shanna Sichwart et al.
Applied and environmental microbiology, 77(4), 1325-1334 (2010-12-21)
The gram-negative facultative chemolithoautotrophic bacterium Ralstonia eutropha strain H16 is known for its narrow carbohydrate utilization range, which limits its use for biotechnological production of polyhydroxyalkanoates and possibly other products from renewable resources. To broaden its substrate utilization range, which
Matthew R Amos et al.
Applied and environmental microbiology, 77(3), 776-785 (2010-12-15)
Extracellular polysaccharide (EPS) is produced by diverse bacterial pathogens and fulfills assorted roles, including providing a structural matrix for biofilm formation and more specific functions in virulence, such as protection against immune defenses. We report here the first investigation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service