Skip to Content
Merck

PGI-RO

Roche

Phosphoglucose Isomerase (PGI)

from yeast

Synonym(s):

glucose-6-phosphate isomerase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
~350 units/mg protein (at 25 °C with fructose-6-P as the substrate.)
Biological source:
yeast
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

yeast

form

suspension

specific activity

~350 units/mg protein (at 25 °C with fructose-6-P as the substrate.)

packaging

pkg of 1 mL (10127396001 [2 mg]), pkg of 1 mL (10128139001 [10 mg])

manufacturer/tradename

Roche

optimum pH

7.0-7.6

storage temp.

2-8°C

General description

Phosphoglucose Isomerase is an enzyme which possesses diagnostic potential when fractionated by cellulose acetate electrophoresis. This cytosolic enzyme plays an important role during glycolysis and glucogenesis. It acts as an tumor-secreted cytokine and autocrine motility factor. It is used as neurotrophic factor, maturation factor and as serine proteinase inhibitor.

Application

Isomerization of ketoses to aldoses.

Biochem/physiol Actions

Phosphoglucose isomerase (PGI) is specific for G6P (Km = 0.7 mM) and fructose-6-phosphate (F6P).

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Preparation Note

Storage conditions (working solution): A solution of PGI in 20 mM Tris, pH 8.1, containing 0.2 mg BSA/ml, is stable approx. 1 week at 2 to 8 °C.

Analysis Note

Contaminants: <0.01% F6P-K, GR, 6-PGDH and PGluM, each, <0.2% β-fructosidase

Other Notes

For life science research only. Not for use in diagnostic procedures.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Aamir Ahmad et al.
Cancer research, 71(9), 3400-3409 (2011-03-11)
Phosphoglucose isomerase/autocrine motility factor (PGI/AMF) plays an important role in glycolysis and gluconeogenesis and is associated with invasion and metastasis of cancer cells. We have previously shown its role in the induction of epithelial-mesenchymal transition (EMT) in breast cancer cells



Global Trade Item Number

SKUGTIN
1012739600104061838250353
1012813900104061838667656