Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
assay
≥97%
form
powder
mol wt
Mr 743.4, Mr 787.4
packaging
pkg of 1 g (10128058001), pkg of 100 mg (10128031001), pkg of 5 g (10240354001), pkg of 500 mg (10128040001)
manufacturer/tradename
Roche
pH
4.0 ± 1.0(10mg/mL)
λmax
260 nm
shipped in
wet ice
storage temp.
2-8°C
SMILES string
[Na+].[Na+].NC(=O)c1ccc[n+](c1)C2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](OP(O)([O-])=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O
InChI
1S/C21H28N7O17P3.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;/q;2*+1/p-2/t10-,11-,13-,14-,15-,16-,20?,21-;;/m1../s1
InChI key
WSDDJLMGYRLUKR-FHQWUTKFSA-L
General description
Application
Preparation Note
10 mg/ml in aqueous solution, contains at least 99% NADP after 4 weeks at 2 to 8 °C
Analysis Note
Absorbance at 340 nm: Imidazol buffer, 100 mM pH 6.7, NADP 2 mM (= 14.87 mg/10 ml) : A340 approximately 0.100.
Absorbance at 360 nm: Aqueous solution of NADP, 10 mg/ml: A360 approximately 0.200-0.250.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Roche | manufacturer/tradename Calbiochem® | manufacturer/tradename Roche | manufacturer/tradename - |
| form powder | form solid | form lyophilized | form solid |
| assay ≥97% | assay ≥92% (enzymatic), ≥95% (UV) | assay 100% (dry weight) | assay ≥95.0% dry basis (enzymatic) |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in - | shipped in wet ice | shipped in - |
| packaging pkg of 1 g (10128058001), pkg of 5 g (10240354001), pkg of 100 mg (10128031001), pkg of 500 mg (10128040001) | packaging - | packaging pkg of 1 g (10127965001), pkg of 5 g (10127973001) | packaging - |
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 10128031001 | 04061838667625 |
| 10128040001 | 04061838667632 |
| 10128058001 | 04061838250377 |
| 10240354001 | 04061838250506 |



