Skip to Content
Merck
All Photos(1)

Key Documents

O9755

Supelco

Oil Red O

BioReagent, suitable for electrophoresis

Synonym(s):

1-([4-(Xylylazo)xylyl]azo)-2-naphthol, 1-[2,5-Dimethyl-4-(2,5-dimethylphenylazo)phenylazo]-2-naphthol, Solvent Red 27

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N4O
CAS Number:
Molecular Weight:
408.49
Colour Index Number:
26125
EC Number:
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

form

powder

technique(s)

electrophoresis: suitable

mp

120 °C (dec.) (lit.)

λmax

359 nm
518 nm (2nd)

suitability

suitable for electrophoresis

SMILES string

Cc1ccc(C)c(c1)\N=N\c2cc(C)c(cc2C)\N=N/c3c(O)ccc4ccccc34

InChI

1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,31H,1-4H3

InChI key

NPGIHFRTRXVWOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oil Red O stain is suitable as a lipid/lipoprotein stain on cellulose acetate. A 0.2% stock solution is prepared in methanol. The stock solution (35 ml) is diluted with 10 mL of 1 M NaOH immediately before staining. The dye begins precipitating during staining. After staining (1 hr), the membrane is destained in methanol:glycerol (1:3). Often cited for use with agarose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingxuan Li et al.
Cardiovascular research, 117(2), 450-461 (2020-01-25)
Eva-1 homologue 1 (Eva1a) is a novel protein involved in the regulation of cardiac remodelling and plaque stability, but little is known about its role in re-endothelialization and the development of atherosclerosis (AS). Thus, in the present study, we aimed
Agarose-gel electrophoresis of lipoproteins.
Opplt, J.J. et al.
CRC Handbook of Electrophoresis, 1, 151-182 (1980)
Metodi V Stankov et al.
Antimicrobial agents and chemotherapy, 54(1), 280-287 (2009-10-07)
Mitochondrial dysfunction as a consequence of mitochondrial DNA (mtDNA) depletion due to therapy with nucleoside analogue reverse transcriptase inhibitors (NRTI) has been proposed as a pathogenic mechanism leading to lipoatrophy in HIV-infected patients. The aim of our study was to
Tomoatsu Ikeya et al.
Proceedings. Biological sciences, 276(1674), 3799-3807 (2009-08-21)
Insulin/IGF-like signalling (IIS) is an evolutionarily conserved pathway that has diverse functions in multi-cellular organisms. Mutations that reduce IIS can have pleiotropic effects on growth, development, metabolic homeostasis, fecundity, stress resistance and lifespan. IIS is also modified by extrinsic factors.
Chang-Hua Zhang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(3-4), 221-229 (2012-12-12)
Gegen Qinlian Decoction (GGQLD) is one of the well-known traditional Chinese medicines. Recently, it was reported that GGQLD had good clinical effects on type 2 diabetes mellitus. However, few studies have confirmed in detail the anti-diabetic activities of GGQLD in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service