Skip to Content
Merck
All Photos(2)

Documents

199664

Sigma-Aldrich

Sudan Black B

certified by the Biological Stain Commission

Synonym(s):

Ceres black BN, Fat Black HB, Solvent Black 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24N6
CAS Number:
Molecular Weight:
456.54
Colour Index Number:
26150
Beilstein:
723248
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

color

dark brown to very dark brown

mp

120-124 °C (lit.)

λmax

415 nm (2nd)
598 nm

ε (extinction coefficient)

≥20000 at 596-603 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC1(C)Nc2cccc3c(ccc(N1)c23)\N=N\c4ccc(\N=N\c5ccccc5)c6ccccc46

InChI

1S/C29H24N6/c1-29(2)30-26-14-8-13-22-25(17-18-27(31-29)28(22)26)35-34-24-16-15-23(20-11-6-7-12-21(20)24)33-32-19-9-4-3-5-10-19/h3-18,30-31H,1-2H3/b33-32+,35-34+

InChI key

YCUVUDODLRLVIC-VPHDGDOJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sudan Black B has been used in brain BLAQ (Block Lipids and Aldehyde Quench) technique and is used in lipid detection.

Biochem/physiol Actions

Sudan Black B has been used to stain phospholipids and intracellular lipids. Staining pattern is almost similar to myeloperoxidase reaction, with positive staining of granulocytic cells, eosinophils and weak monocytic staining.It doesn′t stain lymphocytes. Sudan Black B can be used to stain older blood or bone marrow smears, but the stain slightly fades over time. It is considered to be a most sensitive and specific reagent for lipid staining. Sudan Black B stains phospholipids gray and neutral fats blue-black.

Suitability

Certified as a fat staining using the Churukian variant of the Lillie-Ashburn supersaturated isopropanol method. Sudan III is substituted for the oil red O in the procedure. Burdon′s stain for bacterial lipid.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetylated Sudan Black B as a More Specific Histochemical Reagent for Lipides
CASSELMAN WGB
Journal of Cell Science, 321- 322 (1954)
H N Subramaniam et al.
Journal of biochemical and biophysical methods, 21(1), 9-16 (1990-06-01)
The selective staining of neutral lipids in Human Amnion cells in tissue culture was achieved using a particular fraction of the lipid dye, Sudan black B and a standardized staining procedure. The fraction, termed SBB-I, was isolated by thin-layer chromatography.
Brain BLAQ: Post-hoc thick-section histochemistry for localizing optogenetic constructs in neurons and their distal terminals
Kupferschmidt DA
Frontiers in Neuroanatomy (2015)
Effects of feeding different lipid sources on hepatic histopathology features and growth traits of broiler chickens
Tufarelli V, et.al.
Acta Histochemica, 117, 780-783 (2015)
John P. Greer
Wintrobe's Clinical Hematology, 1 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service