Skip to Content
Merck
All Photos(1)

Key Documents

19123

Sigma-Aldrich

Nile Red

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

Nile Blue A Oxazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N2O2
CAS Number:
Molecular Weight:
318.37
Beilstein:
279110
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Quality Level

Assay

≥97.0% (HPLC)

form

solid

mp

203-205 °C (lit.)
205-215 °C

λmax

553 nm

fluorescence

λex 530 nm; λem 635 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2N=C3C(Oc2c1)=CC(=O)c4ccccc34

InChI

1S/C20H18N2O2/c1-3-22(4-2)13-9-10-16-18(11-13)24-19-12-17(23)14-7-5-6-8-15(14)20(19)21-16/h5-12H,3-4H2,1-2H3

InChI key

VOFUROIFQGPCGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nile Red is an uncharged hydrophobic molecule and a benzophenoxazone dye. It functions as a fluorescent probe for intracellular lipids and hydrophobic domains of proteins. This dye is fluorescent in all organic solvents. Its fluorescence colors range from golden yellow to deep red.

Application

Nile Red has been used for the staining of intracellular lipid droplets and neutral lipids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an in vitro human liver system for interrogating nonalcoholic steatohepatitis
Feaver RE, et al.
JCI insight, 1(20) (2016)
Three-dimensional label-free imaging and quantification of lipid droplets in live hepatocytes
Kim K, et al.
Scientific Reports, 6, 36815-36815 (2016)
Nile red as a polarity-sensitive fluorescent probe of hydrophobic protein surfaces
Sackett D L and Wolff J
Analytical Biochemistry, 167(2), 228-234 (1987)
Biosynthesis of high yield fatty acids from Chlorella vulgaris NIES-227 under nitrogen starvation stress during heterotrophic cultivation
Shen XF, et al.
Water Research, 81(3), 294-300 (2015)
Resveratrol suppresses T0901317-induced hepatic fat accumulation in mice
Gao M and Liu D
The AAPS Journal, 15, 744-752 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service