Skip to Content
Merck
All Photos(2)

Key Documents

O1391

Sigma-Aldrich

Oil Red O solution

0.5% in isopropanol

Synonym(s):

Solvent Red 27, Sudan Red 5B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N4O
CAS Number:
Molecular Weight:
408.49
EC Number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

concentration

0.5% in isopropanol

technique(s)

microbe id | staining: suitable

color

dark red

εmax

0.20-0.50 at 510-517 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N(\N=C3\c4c(cccc4)C=CC\3=O)c1c(cc(c(c1)C)N=Nc2c(ccc(c2)C)C)C

InChI

1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,29H,1-4H3/b28-27?,30-26-

InChI key

HJAYEODIXUYVIC-LTFAAIOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oil Red O stains lipid materials and imparts red-orange color to the lipids. It also marks vacuoles, particularly observed in ALL-L3 (acute lymphoblastic leukemia) or Burkitt′s lymphoma/leukemia. Oil Red O staining is done on fresh samples, as alcohol fixation removes lipids.

Application

Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.
Oil red O solution has been used to stain neutral lipids and adipocytes and to visualize fat droplets in fibroblasts.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepatic Peroxisome Proliferator-Activated Receptor Gamma Signaling Contributes to Alcohol-Induced Hepatic Steatosis and Inflammation in Mice.
Zhang W, et al.
Alcoholism, Clinical and Experimental Research, 40(5), 988-999 (2016)
Directly reprogramming fibroblasts into adipogenic, neurogenic and hepatogenic differentiation lineages by defined factors.
Wu W, et al.
Experimental and Therapeutic Medicine, 13(6), 2685-2690 (2017)
LWW Doody's all reviewed collection, McClatchey: Clinical Laboratory Medicine
Clinical Laboratory Medicine (2002)
Sulforaphane inhibits mammary adipogenesis by targeting adipose mesenchymal stem cells.
Li Q, et al.
Breast Cancer Research and Treatment, 141(2), 317-324 (2013)
Pablo Sanchez Bosch et al.
Developmental cell, 51(6), 787-803 (2019-11-19)
The use of adult Drosophila melanogaster as a model for hematopoiesis or organismal immunity has been debated. Addressing this question, we identify an extensive reservoir of blood cells (hemocytes) at the respiratory epithelia (tracheal air sacs) of the thorax and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service