Skip to Content
Merck
All Photos(1)

Documents

N4007

Sigma-Aldrich

N-Nitroso-N-methylbutylamine

Synonym(s):

Methylbutylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

form

liquid

density

0.93 g/mL (lit.)

storage temp.

2-8°C

SMILES string

CCCCN(C)N=O

InChI

1S/C5H12N2O/c1-3-4-5-7(2)6-8/h3-5H2,1-2H3

InChI key

PKTSCJXWLVREKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bulk package

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Farrelly et al.
Cancer research, 42(6), 2105-2109 (1982-06-01)
With the use of rat liver preparations, the in vitro microsomal metabolism of methylethylnitrosamine, methyl-n-butylnitrosamine, and methyl(2-phenylethyl)nitrosamine labeled with deuterium in the methyl and alpha-methylene positions has been compared with that of the parent (unlabeled) compounds. All three forms of
K E Appel et al.
Carcinogenesis, 7(4), 659-663 (1986-04-01)
To demonstrate whether there are any pathways of nitrite formation from N-nitrosamines other than reductive denitrosation by cytochrome P-450 we performed the following experiments. An esterified alpha-hydroxylated nitrosamine was incubated in a microsomal system to test if nitrite generation is
W Lijinsky et al.
Journal of cancer research and clinical oncology, 106(3), 171-175 (1983-01-01)
The carcinogenic effectiveness of nitrosomethyl-n-butylamine,-n-hexylamine and -n-heptylamine was compared by administration to F344 rats at various concentrations in drinking water and by gavage in oil. Nitrosomethylbutylamine induced only tumors of the upper gastrointestinal tract, and was very toxic. Nitrosomethylhexylamine was
M Lee et al.
Cancer research, 49(6), 1470-1474 (1989-03-15)
Metabolic activation may be a key step in determining the tissue specificity of carcinogenic nitrosamines. In previous work, we characterized P450IIE1 (an acetone/ethanol-inducible form of cytochrome P-450) as the major enzyme for the metabolic activation of N-nitrosodimethylamine. In this work
Q Huang et al.
Cancer letters, 69(2), 107-116 (1993-04-30)
We studied the metabolism of methyl-n-butyl-nitrosamine (MBN), a carcinogen for the rat esophagus and liver. The 2-, 3- and 4-hydroxy derivatives were identified as new metabolites of MBN. In studies on tissue slices freshly removed from MRC-Wistar rats, MBN metabolism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service