Skip to Content
Merck
All Photos(1)

Key Documents

N7382

Sigma-Aldrich

Nitrosomorpholine

Synonym(s):

NMOR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.25

form

solid

SMILES string

O=NN1CCOCC1

InChI

1S/C4H8N2O2/c7-5-6-1-3-8-4-2-6/h1-4H2

InChI key

ZKXDGKXYMTYWTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Tumor initiator in rodent liver, trachea, nasal cavity, esophagus, kidney, lung, and thyroid.

Packaging

Bulk package

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Lijinsky et al.
Carcinogenesis, 3(8), 911-915 (1982-01-01)
A comparison was made of the carcinogenic effectiveness of some cyclic nitrosamines containing oxygen in the ring. The compounds were administered to F344 rats at approximately equimolar concentrations in drinking water. Nitrosomorpholine, nitroso-1,3-oxazolidine and nitrosotetrahydro-1,3-oxazine were of similar potency and
Pinar Erkekoglu et al.
Toxicology mechanisms and methods, 20(2), 45-52 (2010-01-27)
Nitrites are ubiquitous environmental contaminants present in drinking water and foods. Nitrosamines can be formed endogenously from nitrate and nitrite and secondary amines or may be present in food, tobacco smoke, and drinking water. The major goal of this work
Wolf D Kuhlmann et al.
International journal of experimental pathology, 87(5), 343-359 (2006-09-13)
Adult hepatocytes and liver-cell progenitors play a role in restoring liver tissue after injury. For the study of progenitor cells in liver repair, experimental models included (a) surgical removal of liver tissue by partial hepatectomy; (b) acute injury by carbontetrachloride;
Jutta Groos et al.
Toxicological sciences : an official journal of the Society of Toxicology, 99(2), 446-454 (2007-07-20)
Within the scope of the Rat Liver Foci Bioassay the model carcinogens N-nitrosomorpholine (NNM), 2-acetylaminoflouren (2-AAF), phenobarbital (PB), and clofibrate (CF) were analyzed concerning their potency and dose-response relationship to induce foci of altered hepatocytes (FAHs), which are known to
M Tatsuta et al.
Cancer letters, 119(1), 109-113 (2008-04-01)
The effects of prolonged administration of the diuretic amiloride on hepatocarcinogenesis induced by N-nitrosomorpholine (NNM) and the labeling index of the liver were investigated in male Sprague-Dawley rats. Rats were given drinking water containing NNM for 8 weeks and received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service