Skip to Content
Merck
All Photos(1)

Key Documents

40458

Supelco

1-Nitrosopiperidine solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

N1(CCCCC1)N=O

InChI

1S/C5H10N2O/c8-6-7-4-2-1-3-5-7/h1-5H2

InChI key

UWSDONTXWQOZFN-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of the oxidation of N-aminopiperidine with chloramine
Darwich.Ch, et al.
Kinetics and Catalysis, 50(1), 103-110 (2009)
Kinetics of the oxidation of N-aminopiperidine with chloramine
Darwich.Ch, et al.
Kinetics and Catalysis, 50, 103-110 (2009)
P Levallois et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 38(11), 1013-1019 (2000-10-19)
Urinary excretion of volatile nitrosamines was assessed in 59 non-smokers living in a rural county of Québec, Canada. Water and food intakes were measured by means of a 24-hour recall. Nitrates were analyzed in the tap water of all participants
N-Nitrosamines (15 listings): N-Nitrosopiperidine.
Report on carcinogens : carcinogen profiles, 12, 321-322 (2011-08-24)
Y Yamashita et al.
Mutation research, 348(4), 163-168 (1995-12-01)
N-Nitrosodialkylamines are promutagens and proclastogens, requiring metabolic activation for their actions. Previously, we showed that direct-acting bacterial mutagens can be formed from N-nitrosodialkylamines on exposure to near-UV. We have now found that N-nitrosodialkylamines with near-UV irradiation are clastogenic to Chinese

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service