Skip to Content
Merck
All Photos(1)

Documents

PHR3607

Supelco

N-Nitrosodiisopropylamine (NDIPA)

certified reference material, pharmaceutical secondary standard

Synonym(s):

N-Isopropyl-N-nitrosoisopropylamine, DIPNA, NDIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1466663

form

methanol solution

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

application(s)

pharmaceutical

storage temp.

-10 to -25°C

InChI

1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3

InChI key

AUIKJTGFPFLMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Nitrosodiisopropylamine (NDIPA) pharmaceutical secondary reference standard is used for determining NDIPA impurity in sartan pharmaceutical products using High-Performance Liquid Chromatography-Tandem Mass Spectrometry (HPLC-MS/MS).

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Solvent

Description

    1000 ug/mL solution in methanol

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Carcinogenic Impurities of N-Nitrosamines in Sartan Pharmaceutical Products Marketed in Brazil: Development and Validation of Method Based on High-Performance Liquid Chromatography-Tandem Mass Spectrometry
Monteiro, MA et al.
Journal of Pharmaceutical Sciences, 112, 1305-1314 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service