MIDCLARET
CellVue® Claret Far Red Fluorescent Cell Linker Midi Kit for General Membrane Labeling
Distributed for Phanos Technologies
Synonym(s):
Far red membrane probe kit
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
manufacturer/tradename
Distributed for Phanos Technologies
Quality Level
storage condition
protect from light
technique(s)
flow cytometry: suitable
fluorescence
λex 655 nm; λem 675 nm
application(s)
cell analysis
detection
detection method
fluorometric
shipped in
ambient
storage temp.
room temp
Application
CellVue Claret kit fluoresces in the far red and may be used for general cell membrane labeling including in vitro cell labeling, in vitro lymphocyte proliferation studies,† in vivo cytotoxicity assays,† and also as a second color for monitoring cell-cell membrane transfer.†
Biochem/physiol Actions
The CellVue Claret Fluorescent Cell Linker kit uses a proprietary membrane labeling technology to stably incorporate a far red fluorescent dye with long aliphatic tails (CellVue Claret) into lipid regions of the cell membrane.†
Packaging
This kit provides enough dye for up to 50 samples of 20 million cells each and enough diluent for up to 30 samples of 20 million cells each when using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye. Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.
Linkage
For additional technical details on Fluorescent Cell Linker Dyes including a comprehensive bibliography, please visit PKH and CellVue®; Claret Fluorescent Cell Linker Dyes.
Legal Information
Distributed for Phanos Technologies, Inc.
Patent Information
Patent Information
CellVue is a registered trademark of Phanos Technologies
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Toxicology and applied pharmacology, 259(1), 96-103 (2012-01-03)
The toxicity of carbon nanotubes (CNTs), a highly promising nanomaterial, is similar to that of asbestos because both types of particles have a fibrous shape and are biopersistent. Here, we investigated the characteristics of macrophage receptor with collagenous structure (MARCO)
The Journal of clinical investigation, 120(7), 2486-2496 (2010-06-17)
One of the main unresolved questions in solid organ transplantation is how to establish indefinite graft survival that is free from long-term treatment with immunosuppressive drugs and chronic rejection (i.e., the establishment of tolerance). The failure to achieve this goal
Pediatric blood & cancer, 66(5), e27601-e27601 (2019-01-05)
Sickle cell disease (SCD) is increasingly recognized as a red blood cell disorder modulated by abnormally increased inflammation. We have previously shown that in patients with SCD not on a disease-modifying therapy (hydroxyurea or chronic transfusions), natural killer (NK) cell
Biotechnology and bioengineering, 108(9), 2003-2010 (2011-04-16)
In higher eukaryotes, E2F transcription factors often drive expression of genes necessary for the cell cycle, notably the G1/S phase transition. With conventional transcriptional reporter systems, expression of a reporter gene from an E2F-responsive promoter would allow one to identify
Journal of immunology (Baltimore, Md. : 1950), 187(5), 2112-2121 (2011-07-29)
The analysis of cell types involved in cross-priming of particulate Ag is essential to understand and improve immunotherapies using microparticles. In this study, we show that murine splenic dendritic cells (DCs) as well as macrophages (MΦs) are able to efficiently
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service