Skip to Content
Merck
All Photos(1)

Documents

A2264

Sigma-Aldrich

β-N-Acetylglucosaminidase from Canavalia ensiformis (Jack bean)

ammonium sulfate suspension, ≥10 units/mg protein

Synonym(s):

β-N-Acetyl-D-hexosaminide N-acetylhexosaminohydrolase, β-N-Acetylhexosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Canavalia ensiformis

Quality Level

form

ammonium sulfate suspension

specific activity

≥10 units/mg protein

foreign activity

α- and β-galactosidase, and α-L-fucosidase ≤0.1%
α-mannosidase ≤0.2%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

β -N-acetylglucosaminidase is a lysosomal enzyme used to hydrolyze N-acetyl-β-D-glucosaminides and N-acetyl-β-Dgalactosaminides. It is used in chemoenzymatic synthesis of oligosaccharides based on their effective transglycosylation of β-GlcNAc and β-GalNAcc. It may be a useful tool to study Alzheimer′s Disease . Acetylglucosaminidase from Canavalia ensiformis has been used to study enzymic detachment of biofilms .

Biochem/physiol Actions

This enzyme, sometimes called β-N-acetylhexosaminidase, is reported to liberate terminal β-linked N-acetylglucosamine and N-acetylgalactosamine from a variety of substrates. The activity of β-N-actylglucosaminidase may be determined with the chromogenic substrate p-nitrophenyl-N-acetyl-β-D-glucosaminide. β-N-actylglucosaminidase hydrolyzes the terminal nonreducing N-acetyl-D-hexosamine residues. This enzyme contains two predominant isozymes, Hex A, a heterodimer, and Hex B, a homodimer. N-acetylglucosamine, acetamide, N-2-acetamido-2-deoyglucosylamine, N-acetylnojirimycin, and N-acetyldeoxynojirmycin are known inhibitors.
This enzyme, sometimes called β-N-acetylhexosaminidase, is reported to liberate terminal β-linked N-acetylglucosamine and N-acetylgalactosamine from a variety of substrates.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl N-acetyl-β-D-glucosaminide to p-nitrophenol and N-acetyl-D-glucosamine per min at pH 5.0 at 25 °C.

Physical form

Suspension in 2.5 M (NH4)2SO4, pH 7.0

Analysis Note

At pH 4.0, p-nitrophenyl β-N-acetylgalactosaminide is hydrolyzed at approximately 50% of the rate of hydrolysis of p-nitrophenyl β-N-acetylglucosaminide at pH 5.0.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed A Al-Fattani et al.
Journal of medical microbiology, 55(Pt 8), 999-1008 (2006-07-20)
Matrix material was extracted from biofilms of Candida albicans and Candida tropicalis and analysed chemically. Both preparations contained carbohydrate, protein, hexosamine, phosphorus and uronic acid. However, the major component in C. albicans matrix was glucose (32%), whereas in C. tropicalis
T Berger et al.
Journal of reproduction and fertility, 86(2), 559-565 (1989-07-01)
An assay to determine the binding of pig spermatozoa to the zona pellucida (ZP) of pig oocytes was developed using conditions compatible with in-vitro fertilization of pig eggs and with pig sperm penetration of zona-free hamster ova. These conditions were
Chaeyoung Kim et al.
Neurobiology of aging, 34(1), 275-285 (2012-04-17)
Deposition of β-amyloid (Aβ) as senile plaques and disrupted glucose metabolism are two main characteristics of Alzheimer's disease (AD). It is unknown, however, how these two processes are related in AD. Here we examined the relationship between O-GlcNAcylation, which is
Federica Vernuccio et al.
Recenti progressi in medicina, 103(12), 559-563 (2012-12-22)
Cardiorenal syndrome is a pathophysiological heart and kidney disorder, in which acute or chronic dysfunction of one organ induces a damage in the other. It's a syndrome more and more often encountered in clinical practice and this implies the need
Tyler Mark Pierson et al.
Molecular genetics and metabolism, 108(1), 65-69 (2012-11-20)
A 12 year-old female presented with a seven-year history of progressive muscle weakness, atrophy, tremor and fasciculations. Cognition was normal. Rectal biopsy revealed intracellular storage material and biochemical testing indicated low hexosaminidase activity consistent with juvenile-onset G(M2)-gangliosidosis. Genetic evaluation revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service