Skip to Content
Merck
All Photos(1)

Key Documents

F8435

Sigma-Aldrich

PNGase F from Elizabethkingia meningoseptica

lyophilized powder, recombinant, expressed in E. coli

Synonym(s):

N-Glycosidase F, PNGase F from Chryseobacterium meningosepticum, PNGase F from Flavobacterium meningosepticum, Peptide N-glycosidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

grade

Proteomics Grade

form

lyophilized powder

specific activity

≥1,000 U/mg

shelf life

≥1 weeks at 2‑8 °C (for reconstituted solution)
≥1 yr at -20 °C

mol wt

~36 kDa

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Highly purified material can be used for preparative deglycosylation or for analytical applications in gel, in solution, or on blot membranes. The enzyme can be removed from preparative operations by utilizing its C-teminal 6x histidine fusion tag.
Used to deglycosylate protein.

Biochem/physiol Actions

Cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain.

Packaging

PNGase F was used for deglycosylation of P-glycoprotein in a study to investigate the dual impact of statins on p-glycoprotein and its effect on doxorubicin cytotoxicity in human neuroblastoma cells. It was used to treat a purified protein, mouse cone ultraviolet (MUV) pigment, before use in quantitative immunoblot analysis in a study It is used to deglycosylate N-linked glycoproteins. Highly purified material can be used for preparative deglycosylation or for analytical applications in gel, in solution, or on blot membranes. The enzyme can be removed from preparative operations by utilizing its C-teminal 6x histidine fusion tag.

Unit Definition

One unit will catalyze the release of N-linked oligosaccharides from 1 nanomole of denatured ribonuclease B in one minute at 37°C at pH 7.5 monitored by SDS-PAGE. One Sigma unit of PNGase F activity is equal to 1 IUB milliunit.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Nordgren et al.
Methods in molecular biology (Clifton, N.J.), 2271, 155-167 (2021-04-29)
O-glycosylation is a difficult posttranslational modification to analyze. O-glycans are labile and often cluster making their analysis by LC-MS very challenging. OpeRATOR is an O-glycan specific protease that cleaves the protein backbone N-terminally of glycosylated serine and threonine residues. This
T H Plummer et al.
The Journal of biological chemistry, 259(17), 10700-10704 (1984-09-10)
Endo-beta-N-acetylglucosaminidase F preparations from Flavobacterium meningosepticum have been found to contain peptide:N-glycosidase activity. Only the second activity, designated as peptide:N-glycosidase F, readily cleaves the beta-aspartylglycosylamine linkage of a fetuin triantennary complex glycopeptide, as shown by the isolation of the corresponding
Evelyn Sieczkowski et al.
International journal of cancer, 126(9), 2025-2035 (2009-09-10)
The development of multidrug resistance (MDR) is a major problem during cancer treatment. Drug efflux via ATP-binding cassette (ABC) transporters is the main mechanism responsible for resistance to chemotherapeutics. We have recently observed that statins enhance susceptibility to doxorubicin-induced apoptosis
Tobias Bierig et al.
Frontiers in bioengineering and biotechnology, 8, 618615-618615 (2021-01-08)
2019-nCoV is the causative agent of the serious, still ongoing, worldwide coronavirus disease (COVID-19) pandemic. High quality recombinant virus proteins are required for research related to the development of vaccines and improved assays, and to the general understanding of virus
Lauren L Daniele et al.
Investigative ophthalmology & visual science, 46(6), 2156-2167 (2005-05-26)
To test the hypothesis that Nrl(-)(/)(-) photoreceptors are cones, by comparing them with WT rods and cones using morphological, molecular, histochemical, and electrophysiological criteria. The photoreceptor layer of fixed retinal tissue of 4- to 6-week-old mice was examined in plastic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service