A0810
Endoglycosidase H from Streptomyces plicatus
recombinant, expressed in E. coli, buffered aqueous solution
Synonym(s):
β-N-Acetylglucosaminidase H
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Recommended Products
recombinant
expressed in E. coli
conjugate
(N-linked)
form
buffered aqueous solution
shipped in
wet ice
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Endoglycosidase H or endo-β-N-acetylglucosaminidase H is an glycohydrolase. It is produced by Streptomyces plicatus and other Streptomyces species.
Biochem/physiol Actions
Endoglycosidase H is involved in cleaving the N-linked glycans present between the two N-acetylglucosamine (GlcNAc) residues in the core of the glycan chain in high-mannose sugars.
Unit Definition
One unit will release N-linked oligosaccharides from 1 μmole of denatured ribonuclease B per min at 37 °C at pH 5.5.
Physical form
Solution in 20 mM Tris HCl, pH 7.5, containing 50 mM NaCl, 1 mM EDTA
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
High-Level Expression of Endo-
Freeze H H and Kranz C
Current Protocols in Molecular Biology, 0 17(3) (2010)
High-Level Expression of Endo-
Wang F, et al.
Testing, 10(3) (2015)
Roger S Zou et al.
Aging, 3(10), 968-984 (2011-10-13)
A distinct conformational transition from the α-helix-rich cellular prion protein (PrPC) into its β-sheet-rich pathological isoform (PrPSc) is the hallmark of prion diseases, a group of fatal transmissible encephalopathies that includes spontaneous and acquired forms. Recently, a PrPSc-like intermediate form
T Tai et al.
The Journal of biological chemistry, 250(21), 8569-8575 (1975-11-10)
Heterogeneities of the two ovalbumin glycopeptides, (Man)5(GlcNAc)2Asn and (Man)6(GlcNAc)2Asn, were revealed by borate paper electrophoresis of oligosaccharide alcohols obtained from the glycopeptides by endo-beta-N-acetylglucosaminidase H digestion and NaB3H4 reduction. The structures of the major components of the oligosaccharides were determined
Isabelle Chantret et al.
PloS one, 5(7), e11734-e11734 (2010-07-30)
During mammalian protein N-glycosylation, 20% of all dolichol-linked oligosaccharides (LLO) appear as free oligosaccharides (fOS) bearing the di-N-acetylchitobiose (fOSGN2), or a single N-acetylglucosamine (fOSGN), moiety at their reducing termini. After sequential trimming by cytosolic endo beta-N-acetylglucosaminidase (ENGase) and Man2c1 mannosidase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service