Skip to Content
Merck
All Photos(4)

Key Documents

M7133

Sigma-Aldrich

4-Methylumbelliferyl sulfate potassium salt

sulfatase substrate

Synonym(s):

Potassium 4-methylumbelliferyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7KO6S
CAS Number:
Molecular Weight:
294.32
Beilstein:
3803203
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥99% (HPLC)

form

powder

solubility

water: 5 mg/mL, clear, colorless

fluorescence

λex 334 nm; λem 370 nm (pH 10.4)
λex 360 nm; λem 449 nm (Reaction products)

storage temp.

−20°C

SMILES string

[K+].CC1=CC(=O)Oc2cc(OS([O-])(=O)=O)ccc12

InChI

1S/C10H8O6S.K/c1-6-4-10(11)15-9-5-7(2-3-8(6)9)16-17(12,13)14;/h2-5H,1H3,(H,12,13,14);/q;+1/p-1

InChI key

CSOCSPXOODWGLJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylumbelliferyl sulfate (4MUS), a general substrate, is a polar metabolite of 4-methylumbelliferone (4MU).

Application

4-Methylumbelliferyl sulfate potassium salt has been used as a substrate in coupled fluorescence sulfotransferase assay to regenerate 3′-phosphoadenosine 5′-phosphosulfate (PAPS) from 3′-phosphoadenosine 5′-phosphate for cerebroside sulfotransferase (CST) enzyme activity measurements. It has also been used as a substrate in arylsulfatase assay/4-methylumbelliferyl sulfate (4-MUS) assay to test the enzymatic activity of the protein-bead complex.

Biochem/physiol Actions

4-Methylumbelliferyl sulfate (4MUS) is capable of undergoing desulfation and involve in futile cycling with 4MU. It can be utilized to measure the arylsulfatase activity of several sulfatases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O P van Diggelen et al.
Journal of inherited metabolic disease, 12(3), 273-280 (1989-01-01)
Arylsulphatase C (ASC) activity in leukocytes and fibroblasts measured with 4-methylumbelliferylsulphate, is caused by at least two genetically different sulphatases. One of these is steroid sulphatase (STS). Depending on the substrate concentration, about 10-50% of the ASC activity in leukocytes
A M Hassen et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(7), 792-798 (1996-07-01)
The uptake of estrone sulfate (E1S; 1 to 400 microM), harmol sulfate (HS; 5 to 900 microM), and 4-methylumbelliferyl sulfate (4MUS; 5 to 1000 microM) was investigated in isolated rat hepatocytes in the presence or absence of inhibitors. Uptake of
M Chiba et al.
The Journal of pharmacology and experimental therapeutics, 266(2), 492-499 (1993-08-01)
4-Methylumbelliferyl sulfate (4MUS), a polar metabolite of 4-methylumbelliferone (4MU), is known to undergo desulfation and participate in futile cycling with 4MU. Unusual parabolic or increasing profiles of the steady-state extraction ratio (Ess) of 4MUS with respect to concentration in rat
Renhong Tang et al.
The Journal of biological chemistry, 284(32), 21505-21514 (2009-06-13)
Sulf-1 and Sulf-2 are novel extracellular sulfatases that act on internal glucosamine 6-O-sulfate modifications within heparan sulfate proteoglycans and regulate their interactions with various signaling molecules, including Wnt ligands. Although the Sulfs are multidomain proteins, there is limited information available
Anthony D Verderosa et al.
Scientific reports, 11(1), 1569-1569 (2021-01-17)
Antibiotics are failing fast, and the development pipeline remains alarmingly dry. New drug research and development is being urged by world health officials, with new antibacterials against multidrug-resistant Gram-negative pathogens as the highest priority. Antivirulence drugs, which inhibit bacterial pathogenicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service