Skip to Content
Merck
All Photos(4)

Key Documents

M7008

Sigma-Aldrich

4-Methylumbelliferyl-β-D-xylopyranoside

β-xylosidase substrate, fluorogenic, ≥98% (HPLC), powder

Synonym(s):

4-Methylumbelliferyl β-D-xyloside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O7
CAS Number:
Molecular Weight:
308.28
Beilstein:
39934
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

Assay

≥98% (HPLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)ccc12

InChI

1S/C15H16O7/c1-7-4-12(17)22-11-5-8(2-3-9(7)11)21-15-14(19)13(18)10(16)6-20-15/h2-5,10,13-16,18-19H,6H2,1H3/t10-,13+,14-,15+/m1/s1

InChI key

JWIYLOHVJDJZOQ-KAOXEZKKSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

A substrate for β-xylosidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shibata et al.
The Journal of biological chemistry, 270(23), 13794-13798 (1995-06-09)
4-Methylumbelliferyl-beta-D-xyloside (Xyl-MU) was added to the medium of cultured human skin fibroblasts. After incubation, the culture medium was pooled, and the Xyl-MU-induced oligosaccharides in the medium were purified by gel filtration chromatography. A novel Xyl-MU derivative was obtained, in addition
K Takagaki et al.
Journal of biochemistry, 119(4), 697-702 (1996-04-01)
Human skin fibroblasts were incubated in the presence of a fluorogenic xyloside, 4-methyl-umbelliferyl-beta-D-xyloside (Xyl-MU), then the cultured medium was recovered, concentrated with a lyophilizer, and dialyzed against distilled water. The structures of the Xyl-MU derivatives purified from the dialyzable fraction
Loontiens, F.G. and DeBruyne, C.K.
Naturwissenschaften, 52, 661-661 (1965)
H H Freeze et al.
The Journal of biological chemistry, 268(3), 1618-1627 (1993-01-25)
beta-D-Xylosides are often used to competitively inhibit proteoglycan synthesis by serving as primers for free glycosaminoglycan (GAG) chain assembly. Quite unexpectedly, we found that when human melanoma cells and Chinese hamster ovary cells are labeled with [3H] galactose in the
J W Burgess et al.
Biochemistry, 38(2), 524-531 (1999-01-15)
We have investigated the association of apolipoprotein E (apoE) with the HepG2 cell surface (i.e. plasma membrane and extracellular matrix) using domain specific monoclonal antibodies against apoE. Growth in beta-D-xyloside decreased the incorporation of 35S into glycosaminoglycans by 31% and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service