Skip to Content
Merck
All Photos(4)

Key Documents

M2133

Sigma-Aldrich

4-Methylumbelliferyl N-acetyl-β-D-glucosaminide

β-N-acetylhexosaminidase substrate, fluorogenic, ≥98% (TLC), powder

Synonym(s):

4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-gluco­pyrano­side

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
EC Number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, β-N-acetylhexosaminidase substrate

Assay

≥98% (TLC)

form

powder

solubility

DMF: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16-,17-,18?/m1/s1

InChI key

QCTHLCFVVACBSA-XWEABGILSA-N

General description

4-Methylumbelliferyl N-acetyl-β-D-glucosaminide (4-MUF-NAG) contains chitin monomer, N-acetyl-D-glucosamine (NAG) and a 4-methylumbelliferone (7-hydroxy-4-methylcoumarin) (4-MUF) moiety.

Application

4-Methylumbelliferyl N-acetyl-β-D-glucosaminide has been used as a substrate for assaying
  • β-N-acetylglucosaminidase (NAGase) from soil samples
  • chitinase from Fusarium oxysporum using fluorimetric assay
  • β-N-acetylhexosaminidases from Arabidopsis thaliana roots and leaves samples

Biochem/physiol Actions

4-Methylumbelliferyl N-acetyl-β-D-glucosaminide (4-MUF-NAG) is a synthetic fluorogenic substrate for N-acetyl-β-D-glucosaminidase (NAGase). The enzyme functionality is assessed by its ability to hydrolyze the N-acetyl-D-glucosamine (NAG) residue in 4-MUF-NAG. 4-MUF-NAG is also a substrate for Chitinase and N-acetylhexosaminidase.

Substrates

Fluorogenic substrate for β-N-acetylhexosaminidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of Chitinase Activity in Fusarium oxysporum
Ruiz-Roldan, Carmen and Roncero, M Isabel G
Bio-protocol, 6(15), 98-102 (2016)
beta-N-Acetylhexosaminidases HEXO1 and HEXO3 are responsible for the formation of paucimannosidic N-glycans in Arabidopsis thaliana
Liebminger E, et al.
Test, 286(12), 10793-10802 (2011)
Considerations in the storage of soil samples for enzyme activity analysis
Peoples MS and Koide RT
Applied soil ecology : a section of Agriculture, Ecosystems & Environment, 62(1), 98-102 (2012)
Purification and characterization of beta-N-acetylhexosaminidase I2 from human liver
Dewji NN, et al.
The Biochemical Journal, 234(1), 157-162 (1986)
Li Ou et al.
Gene therapy, 27(5), 226-236 (2020-01-04)
The GM2-gangliosidoses are neurological diseases causing premature death, thus developing effective treatment protocols is urgent. GM2-gangliosidoses result from deficiency of a lysosomal enzyme β-hexosaminidase (Hex) and subsequent accumulation of GM2 gangliosides. Genetic changes in HEXA, encoding the Hex α subunit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service