Skip to Content
Merck
All Photos(1)

Key Documents

M6908

Sigma-Aldrich

Myeloperoxidase from human leukocytes

lyophilized powder, ≥50 units/mg protein

Synonym(s):

MPO, Peroxidase, myelo

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human leucocytes

Quality Level

form

lyophilized powder

specific activity

≥50 units/mg protein

solubility

H2O: soluble
aqueous buffers such as 50 mM sodium acetate: soluble (Freezing of the enzyme in solution may result in a substantial loss of activity.)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Gene Information

human ... MPO(4353)

Looking for similar products? Visit Product Comparison Guide

General description

Myeloperoxidase (MPO) is encoded by the gene mapped to human chromosome 17q22-24. The encoded protein is a leukocyte-derived enzyme and is an integral constituent of the innate immune response. MPO belongs to the heme peroxidase superfamily. MPO is a heme protein containing iron and is highly expressed in leukocytes, neutrophils, monocytes and some subtypes of tissue macrophages.

Application

Myeloperoxidase (MPO) from human leukocytes has been used in MPO assay.
Myeloperoxidase from human leukocytes has been used:
  • as a standard to quantify the neutrophil (polymorphonuclear cell [PMN]) infiltration in lung tissue
  • as a neutrophil extracelullar trap (NET) component to stimulate 106 dendritic cells or 106 macrophages
  • as a standard in myeloperoxidase (MPO) assay to measure pancreatic MPO

Biochem/physiol Actions

Myeloperoxidase (MPO) has a crucial role to play in destruction of various microorganisms and foreign cells, such as bacteria, fungi, viruses, red cells and malignant and nonmalignant nucleated cells. MPO catalyzes the production of number of reactive oxidant species (ROS) that influence tissue damage during inflammation. MPO and its downstream inflammatory pathways function as a potent therapeutic target for prophylaxis of atherosclerotic cardiovascular disease. MPO catalyze the synthesis of polychlorinated dibenzo(p)dioxins and furans (PCDD/F) from precursor such as chlorophenols in presence of hydrogen peroxide (H2O2).

Other Notes

Lysosomal heme-containing protein.

Unit Definition

One unit of myeloperoxidase will catalyze the consumption of one micromole of hydrogen peroxide and the production of 1/4 micromole of tetraguaiacol per minute at pH 7.0 and 25 °C

Physical form

Lyophilized from 50 mM sodium acetate buffer, pH 6.0, 0.1 M sodium chloride

Analysis Note

Source material tested and found negative for antibody to HIV and for HBSAG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avinash Khandagale et al.
Biology open, 7(7) (2018-06-28)
Patients with inflammatory bowel disease (IBD) are susceptible to thromboembolism. Interestingly, IBD occurs less frequently in patients with inherited bleeding disorders. Therefore, we analyzed whether F9-deficiency is protective against the onset of acute colitis in a genetic hemophilia B mouse
Michael S Runyon et al.
Thrombosis journal, 8, 3-3 (2010-02-26)
Experimental models of pulmonary embolism (PE) that produce pulmonary hypertension (PH) employ many different methods of inducing acute pulmonary occlusion. Many of these models induce PE with intravenous injection of exogenous impervious objects that may not completely reproduce the physiological
The anti-inflammatory modulatory role of Solidago chilensis Meyen in the murine model of the air pouch.
Liz R
The Journal of Pharmacy and Pharmacology, 60(4), 515-521 (2008)
Bronchial inflammation in acute bacterial exacerbations of chronic bronchitis: the role of leukotriene B4.
Crooks SW
The European Respiratory Journal, 15(2), 274-280 (2000)
Cytokine Kinetics and Other Host Factors in Response to Pneumococcal Pulmonary Infection in Mice
Bergeron Y, et al.
Clinical Microbiology Reviews, 66(3), 912-922 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service