Skip to Content
Merck
All Photos(3)

Key Documents

M6524

Sigma-Aldrich

Methoxamine hydrochloride

Synonym(s):

α-(1-Aminoethyl)-2,5-dimethoxybenzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

storage temp.

2-8°C

SMILES string

Cl[H].COc1ccc(OC)c(c1)C(O)C(C)N

InChI

1S/C11H17NO3.ClH/c1-7(12)11(13)9-6-8(14-2)4-5-10(9)15-3;/h4-7,11,13H,12H2,1-3H3;1H

InChI key

YGRFXPCHZBRUKP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α1-adrenoceptor agonist.

Features and Benefits

This compound is featured on the α1-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven Francque et al.
Laboratory investigation; a journal of technical methods and pathology, 92(10), 1428-1439 (2012-08-15)
Non-alcoholic fatty liver disease can progress to steatohepatitis and fibrosis, and is also associated with impaired liver regeneration. The pathophysiology remains elusive. We recently showed that severe steatosis is associated with an increase in portal pressure, suggesting liver flow impairment.
Robert H Thiele et al.
Anesthesia and analgesia, 113(2), 284-296 (2011-04-27)
Phenylephrine and methoxamine are direct-acting, predominantly α(1) adrenergic receptor (AR) agonists. To better understand their physiologic effects, we screened 463 articles on the basis of PubMed searches of "methoxamine" and "phenylephrine" (limited to human, randomized studies published in English), as
R H Lee et al.
Journal of neurophysiology, 81(5), 2164-2174 (1999-05-13)
Enhancement of bistability in spinal motoneurons in vivo by the noradrenergic alpha1 agonist methoxamine. Like many types of motoneurons, spinal motoneurons in the adult mammal can exhibit bistable behavior. This means that short periods of excitatory input can initiate long
Hideko Inaba et al.
Circulation journal : official journal of the Japanese Circulation Society, 75(1), 89-93 (2010-12-09)
Anesthesia sometimes suppresses ventricular tachyarrhythmias (VT) resistant to conventional pharmacological treatment. To know (1) whether deep anesthesia inhibits abnormal repolarization-related VT and (2) if α2-adrenoreceptor (AR) agonistic action is associated with the antiarrhythmic effect of anesthetics, the incidence of VT
Manuel Hernández-Guerra et al.
Hepatology (Baltimore, Md.), 57(4), 1564-1574 (2012-11-24)
Chronic intermittent hypoxia (CIH) occurs with obstructive sleep apnea syndrome (OSAS) and provokes systemic endothelial dysfunction, which is associated with oxidative stress and low nitric oxide (NO) bioavailability. Cirrhotic livers exhibit intrahepatic endothelial dysfunction, which is characterized by an impaired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service