Skip to Content
Merck
All Photos(1)

Key Documents

15238

Supelco

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane

for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)

Synonym(s):

Silylating mixture V, BSTFA + TMCS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥98.5% (excluding chlorotrimethylsilane, GC)
99% (excluding TMCS)

form

liquid

quality

LiChropur
contains 1% TMCS

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.384 (lit.)

bp

45-50 °C/14 mmHg (lit.)

density

0.97 g/mL (lit.)

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Learn more in the Product Information

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Other Notes

The position of the silyl groups has not been unequivocally established. It has been referred to as both N,N and N,O.
Silylating mixture used for the silylation of various compounds

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.1 °F - closed cup

Flash Point(C)

6.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
W.M. Hammargren et al.
Analytica Chimica Acta, 247, 201-201 (1991)
DNA oxidation matters: the HPLC-electrochemical detection assay of 8-oxo-deoxyguanosine and 8-oxo-guanine.
Helbock H
Proceedings of the National Academy of Sciences of the USA, 95 (1), 288-293 (1998)
Metabolism of arachidonate through NADPH-dependent oxygenase of renal cortex.
Proceedings of the National Academy of Sciences of the USA, 78 (12), 7375-7378 (1981)
W. Greenway et al.
Journal of Chromatography A, 519, 145-145 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service