Skip to Content
Merck
All Photos(1)

Documents

M5775

Sigma-Aldrich

Minimum Essential Medium Eagle

With Hanks′ salts and sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, With Hanks′ salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: yes
Hanks’ salts (2% CO2): yes
L-glutamine: no
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Modification of the original MEM formulation. Earle′s salts have been replaced with Hanks′ salts. Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.

Reconstitution

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maruša Bizjak et al.
Biochemical and biophysical research communications, 515(2), 296-302 (2019-05-31)
Metabolic pathways of cancer cells depend on the concentrations of nutrients in their micro-environment as well as on the cell-to-cell interactions. Here we examined the effects of glucose, pyruvate and glutamine on the sensitivity of MDA-MB-231 cells to metabolic drug metformin
Simona Giunta et al.
The Journal of cell biology, 190(2), 197-207 (2010-07-28)
The signaling cascade initiated in response to DNA double-strand breaks (DSBs) has been extensively investigated in interphase cells. Here, we show that mitotic cells treated with DSB-inducing agents activate a "primary" DNA damage response (DDR) comprised of early signaling events
Pasqualino de Antonellis et al.
PloS one, 6(9), e24584-e24584 (2011-09-21)
Through negative regulation of gene expression, microRNAs (miRNAs) can function as oncosuppressors in cancers, and can themselves show altered expression in various tumor types. Here, we have investigated medulloblastoma tumors (MBs), which arise from an early impairment of developmental processes
Barbara Zonta et al.
Neuron, 69(5), 945-956 (2011-03-09)
The axon initial segment (AIS) is critical for the initiation and propagation of action potentials. Assembly of the AIS requires interactions between scaffolding molecules and voltage-gated sodium channels, but the molecular mechanisms that stabilize the AIS are poorly understood. The
Carlos Mendoza-Palomares et al.
Eukaryotic cell, 7(4), 684-697 (2008-02-19)
Cysteine proteases have been shown to be essential virulence factors and drug targets in trypanosomatids and an attractive antidisease vaccine candidate for Trypanosoma congolense. Here, we describe an important amplification of genes encoding cathepsin B-like proteases unique to T. congolense.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service