Skip to Content
Merck
All Photos(2)

Key Documents

M3281

Sigma-Aldrich

α-Methyl-DL-tyrosine methyl ester hydrochloride

≥95% (HPLC)

Synonym(s):

α-MT methyl ester hydrochloride, 2-Methyl-4-hydroxy-DL-phenylalanine methyl ester hydrochloride, AMPT methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2C(CH3)(NH2)CO2CH3 · HCl
CAS Number:
Molecular Weight:
245.70
Beilstein:
5163031
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

mp

192 °C (dec.) (lit.)

solubility

DMSO: ≥20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl[H].COC(=O)C(C)(N)Cc1ccc(O)cc1

InChI

1S/C11H15NO3.ClH/c1-11(12,10(14)15-2)7-8-3-5-9(13)6-4-8;/h3-6,13H,7,12H2,1-2H3;1H

InChI key

OOVDEPZODSXAMU-UHFFFAOYSA-N

Gene Information

human ... TH(7054)

Looking for similar products? Visit Product Comparison Guide

Application

α-Methyl-DL-tyrosine methyl ester hydrochloride has been used to evaluate norepinephrine turnover.

Biochem/physiol Actions

α-Methyl-DL-tyrosine (α-MT) prevents hyperactivity, sniffing-licking-gnawing syndrome, anorexia and ameliorates certain effects of amphetamine. It inhibits dopamine production and prevents apoptosis stimulated by mutant α-synuclein.
Tyrosine hydroxylase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dopamine-dependent neurotoxicity of alpha-synuclein: a mechanism for selective neurodegeneration in Parkinson disease
Xu J, et al.
Nature Medicine, 8(6), 600-600 (2002)
Central sympathetic innervations to visceral and subcutaneous white adipose tissue
Nguyen, Ngoc Ly T and Randall, Jessica and Banfield, Bruce W and Bartness, Timoth NLT, et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 306(6), R375-R386 (2014)
Antiamphetamine effects following inhibition of tyrosine hydroxylase
Weissman A
Journal of Pharmacology and Experimental Therapeutics, 151(3), 339-352 (1966)
R H Migliorini et al.
The American journal of physiology, 272(2 Pt 2), R656-R661 (1997-02-01)
The effect of prolonged fasting on sympathetic activity was examined in rat white adipose tissue (WAT) and, for comparison purposes, in interscapular brown adipose tissue (IBAT). Preliminary experiments showed that 6-hydroxydopamine or tyramine administration to fed animals produced similar reductions
G H Paalzow
The Journal of pharmacology and experimental therapeutics, 263(2), 470-479 (1992-11-01)
The present study shows that during the time course of the action of single doses, L-dopa induces multiphasic opposing effects on pain, recorded as vocalization during the presentation of electrical stimulation applied to the tail of normal rats. This indicates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service