Skip to Content
Merck
All Photos(4)

Key Documents

M2272

Sigma-Aldrich

Melittin from honey bee venom

≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H229N39O31
CAS Number:
Molecular Weight:
2846.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

animal (Apis mellifera)

Quality Level

Assay

≥85% (HPLC)

form

lyophilized powder

mol wt

~_2.8 kDa

solubility

water: 5.00-5.20 mg/mL, clear, colorless to faintly yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

foreign activity

Phospholipase A2 ≤0.5%

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CN)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O

InChI

1S/C131H229N39O31/c1-23-71(16)102(163-97(176)60-135)122(194)146-62-98(177)148-74(19)109(181)164-100(69(12)13)124(196)160-88(55-65(4)5)116(188)155-84(41-30-33-51-134)115(187)165-101(70(14)15)125(197)161-90(57-67(8)9)118(190)168-106(77(22)173)128(200)169-105(76(21)172)123(195)147-63-99(178)150-92(58-68(10)11)129(201)170-54-36-44-94(170)121(193)149-75(20)108(180)158-89(56-66(6)7)117(189)166-104(73(18)25-3)127(199)162-93(64-171)120(192)159-91(59-78-61-145-80-38-27-26-37-79(78)80)119(191)167-103(72(17)24-2)126(198)157-83(40-29-32-50-133)111(183)154-85(42-34-52-143-130(139)140)112(184)152-82(39-28-31-49-132)110(182)153-86(43-35-53-144-131(141)142)113(185)156-87(46-48-96(137)175)114(186)151-81(107(138)179)45-47-95(136)174/h26-27,37-38,61,65-77,81-94,100-106,145,171-173H,23-25,28-36,39-60,62-64,132-135H2,1-22H3,(H2,136,174)(H2,137,175)(H2,138,179)(H,146,194)(H,147,195)(H,148,177)(H,149,193)(H,150,178)(H,151,186)(H,152,184)(H,153,182)(H,154,183)(H,155,188)(H,156,185)(H,157,198)(H,158,180)(H,159,192)(H,160,196)(H,161,197)(H,162,199)(H,163,176)(H,164,181)(H,165,187)(H,166,189)(H,167,191)(H,168,190)(H,169,200)(H4,139,140,143)(H4,141,142,144)/t71-,72-,73-,74-,75-,76+,77+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,100-,101-,102-,103-,104-,105-,106-/m0/s1

InChI key

VDXZNPDIRNWWCW-JFTDCZMZSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH2

General description

Chemical structure: peptide
Melittin is hydrophobic in nature except for a region with Lys-Arg-Lys-Arg sequence near C-terminal end. This structural characteristic makes melittin a highly surface-active and a powerful, direct haemolytic agent.† The encoded protein containins 26 amino acids. Monomeric form of melittin has a molecular weight of 2,840 Daltons and tetrameric form has molecular weight of approximately 12,500 Daltons.

Application

Melittin from honey bee venom has been used:
  • In 3-(4, 5-dimethyl thiazol-2-yl)-2,5diphenyl tetrazolium bromide (MTT) assay to determine its cytotoxicity effect on the growth of human cell lines.
  • To study the anti-microbial activity of melittin on the growth of Borrelia burgdorferi in in vitro conditions.
  • As a positive control in hemolysis assay and as a cytotoxic agent against HeLa cells.

Biochem/physiol Actions

Binds calmodulin in a Ca2+-dependent manner; inhibits Na+-K+-ATPase.
Melittin acts as an anti-coagulating protein by increasing the time of blood clotting in vitro. Melittin inhibits the activity of S100 calcium-binding protein B (S100B) and plays a vital role in Epilepsy treatment.

Other Notes

The principle hemolytic component of honeybee venom.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ipsita Saha et al.
Viruses, 13(10) (2021-10-27)
Immature HIV virions harbor a lattice of Gag molecules with significant ordering in CA-NTD, CA-CTD and SP1 regions. This ordering plays a major role during HIV maturation. To test the condition in which the Gag lattice forms in vivo, we
The antimicrobial agent melittin exhibits powerful in vitro inhibitory effects on the Lyme disease spirochete.
Lubke LL and Garon CF
Clinical Infectious Diseases, suppl 1, S48-S51 (1997)
Selective cytotoxicity Assay in anticancer drug of Melittin Isolated from
Bee Venom (Apis cerana indica) to several human cell lines: HeLa,
WiDr and Vero.
Plasay M
Journal of Chemical and Pharmaceutical Sciences , 9:4, 2674-2676 (2016)
Structural and Dynamic Insights into S100B Protein Activity Inhibition by Melittin for the Treatment of Epilepsy
Verma N
International Journal of Computer Applications, 55-60 (2013)
Luís Moll et al.
Frontiers in microbiology, 12, 753874-753874 (2021-11-26)
Xylella fastidiosa is a plant pathogen that was recently introduced in Europe and is causing havoc to its agriculture. This Gram-negative bacterium invades the host xylem, multiplies, and forms biofilm occluding the vessels and killing its host. In spite of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service