Skip to Content
Merck
All Photos(2)

Key Documents

P9279

Sigma-Aldrich

Phospholipase A2 from honey bee venom (Apis mellifera)

salt-free, lyophilized powder, 600-2400 units/mg protein

Synonym(s):

Lecithinase A, PLA2, Phosphatidylcholine 2-acylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

form

salt-free, lyophilized powder

Quality Level

specific activity

600-2400 units/mg protein

mol wt

14.5 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

honey bee ... Pla2(406141)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phospholipase A2 from honey bee venom (Apis mellifera) is a calcium-dependent lipolytic enzyme.[1]

Application

Phospholipase A2 from honey bee venom (Apis mellifera) has been used
  • to determine renal proximal tubular segments (PTS) viability during oxygenation and hypoxia-reoxygenation[2]
  • in the treatment of photosystem-II (PSII) dimer complex of Synechocystis to study changes in oxygen-evolving activity[3]
  • as a standard in phospholipase activity assay[4]

Biochem/physiol Actions

Hydrolyzes the β-ester bond of zwitterionic glycerophospholipids. Preferred substrates are phosphatidylcholine, phosphatidylethanolamine, and their plasmalogen analogues. Phosphatidylinositol and phosphatidylserine are also hydrolyzed. Aggressively attacks phospholipids in membranes of intact cells.

Unit Definition

One unit will hydrolyze 1.0 μmole of soybean L-α-phosphatidylcholine to L-α-lysophosphatidylcholine and a fatty acid per min at pH 8.9 at 25 °C.

Analysis Note

Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milleporin-1, a new phospholipase A2 active protein from the fire coral Millepora platyphylla nematocysts.
Radwan FF and Aboul-Dahab HM
Comparative Biochemistry and Physiology. Toxicology & Pharmacology : CBP, 139(4), 267-272 (2004)
Lipids in oxygen-evolving photosystem II complexes of cyanobacteria and higher plants.
Sakurai I
Journal of Biochemistry, 140(2), 201-209 (2006)
Third calcium ion found in an inhibitor-bound phospholipase A2.
Sekar K
Acta Crystallographica Section D, Biological Crystallography, 62, 392-397 (2006)
Phospholipase A2 activity can protect renal tubules from oxygen deprivation injury.
Zager RA
Proceedings of the National Academy of Sciences of the USA, 90(17), 8297-8301 (1993)
Ming Chen et al.
Scientific reports, 7(1), 15931-15931 (2017-11-23)
Hepatitis C virus (HCV), dengue virus (DENV) and Japanese encephalitis virus (JEV) belong to the family Flaviviridae. Their viral particles have the envelope composed of viral proteins and a lipid bilayer acquired from budding through the endoplasmic reticulum (ER). The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service