Skip to Content
Merck
All Photos(1)

Key Documents

M7391

Sigma-Aldrich

Melittin from honey bee venom

≥65% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H229N39O31
CAS Number:
Molecular Weight:
2846.46
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

form

lyophilized powder

Quality Level

mol wt

~_2.8 kDa

concentration

≥65% (HPLC)

solubility

water: 5.00-5.20 mg/mL, clear, colorless to faintly yellow

foreign activity

Phospholipase A2 ≤2%

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CN)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O

InChI

1S/C131H229N39O31/c1-23-71(16)102(163-97(176)60-135)122(194)146-62-98(177)148-74(19)109(181)164-100(69(12)13)124(196)160-88(55-65(4)5)116(188)155-84(41-30-33-51-134)115(187)165-101(70(14)15)125(197)161-90(57-67(8)9)118(190)168-106(77(22)173)128(200)169-105(76(21)172)123(195)147-63-99(178)150-92(58-68(10)11)129(201)170-54-36-44-94(170)121(193)149-75(20)108(180)158-89(56-66(6)7)117(189)166-104(73(18)25-3)127(199)162-93(64-171)120(192)159-91(59-78-61-145-80-38-27-26-37-79(78)80)119(191)167-103(72(17)24-2)126(198)157-83(40-29-32-50-133)111(183)154-85(42-34-52-143-130(139)140)112(184)152-82(39-28-31-49-132)110(182)153-86(43-35-53-144-131(141)142)113(185)156-87(46-48-96(137)175)114(186)151-81(107(138)179)45-47-95(136)174/h26-27,37-38,61,65-77,81-94,100-106,145,171-173H,23-25,28-36,39-60,62-64,132-135H2,1-22H3,(H2,136,174)(H2,137,175)(H2,138,179)(H,146,194)(H,147,195)(H,148,177)(H,149,193)(H,150,178)(H,151,186)(H,152,184)(H,153,182)(H,154,183)(H,155,188)(H,156,185)(H,157,198)(H,158,180)(H,159,192)(H,160,196)(H,161,197)(H,162,199)(H,163,176)(H,164,181)(H,165,187)(H,166,189)(H,167,191)(H,168,190)(H,169,200)(H4,139,140,143)(H4,141,142,144)/t71-,72-,73-,74-,75-,76+,77+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,100-,101-,102-,103-,104-,105-,106-/m0/s1

InChI key

VDXZNPDIRNWWCW-JFTDCZMZSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH2

General description

Melittin is hydrophobic in nature except for a region with Lys-Arg-Lys-Arg sequence near C-terminal end. This structural characteristic makes melittin a highly surface-active and a powerful, direct haemolytic agent. The encoded protein contains 26 amino acids. Monomeric form of melittin has a molecular weight of 2,840Da and tetrameric form has molecular weight of approximately 12,500Da.

Application

Melittin from honey bee venom has been used to study the role of melittin in allergic reactions.

Biochem/physiol Actions

Binds calmodulin in a Ca2+-dependent manner; inhibits Na+-K+-ATPase.
Melittin acts as an anti-coagulating protein by increasing the time of blood clotting in vitro. Melittin inhibits the activity of S100 calcium-binding protein B (S100B) and plays a vital role in Epilepsy treatment. Melittin retard cathepsin S-induced invasion, proliferation and angiogenesis via inhibition of the vascular endothelial growth factor A (VEGF-A) /VEGF receptor 2 (VEGFR-2)/ mitogen-activated protein kinase 1 (MEK1)/ extracellular signal-regulated kinase 1/2 (ERK1/2) pathway in human MHCC97-H cells (liver cancer cells). Melittin has therapeutic application in various inflammatory diseases such as skin inflammation, neuroinflammation, atherosclerosis, arthritis and liver inflammation. Accumulated melittin degrades phospholipid packing in the cell membrane and causes cell lysis and cell death.

Other Notes

The principle hemolytic component of honeybee venom.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anti-Inflammatory Applications of Melittin, a Major Component of Bee Venom: Detailed Mechanism of Action and Adverse Effects.
Molecules (Basel), 21(5) (2016)
Structural and Dynamic Insights into S100B Protein Activity Inhibition by Melittin for the Treatment of Epilepsy
Verma N
International Journal of Computer Applications, 55-60 (2013)
Application of bee venom and its main constituent melittin for cancer treatment.
Application of bee venom and its main constituent melittin for cancer treatment.
Liu CC
Cancer Chemotherapy and Pharmacology, 78(6), 1113-1130 (2016)
Honey Bee Venom (Apis mellifera) Contains Anticoagulation Factors and Increases the Blood-clotting Time.
Zolfagharian H
Journal of pharmacopuncture, 18(4), 7-11 (2015)
Thomas Gutsmann et al.
Antimicrobial agents and chemotherapy, 54(9), 3817-3824 (2010-07-08)
Systemic bacterial infections are associated with high mortality. The access of bacteria or constituents thereof to systemic circulation induces the massive release of immunomodulatory mediators, ultimately causing tissue hypoperfusion and multiple-organ failure despite adequate antibiotic treatment. Lipid A, the "endotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service