Skip to Content
Merck
All Photos(2)

Key Documents

M1390

Sigma-Aldrich

3,3′-Methylene-bis(4-hydroxycoumarin)

Synonym(s):

Bishydroxycoumarin, Dicoumarol, Dicumarol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12O6
CAS Number:
Molecular Weight:
336.29
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.47

biological source

synthetic (organic)

Assay

≥98% (TLC)

form

powder

mp

290-292 °C (lit.)

solubility

pyridine: 50 mg/mL, clear, faintly yellow to brownish-yellow

SMILES string

OC1=C(CC2=C(O)c3ccccc3OC2=O)C(=O)Oc4ccccc14

InChI

1S/C19H12O6/c20-16-10-5-1-3-7-14(10)24-18(22)12(16)9-13-17(21)11-6-2-4-8-15(11)25-19(13)23/h1-8,20-21H,9H2

InChI key

DOBMPNYZJYQDGZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prototype of the 4-hydroxycoumarin class of anticoagulants, which act as vitamin K antagonists, preventing formation of prothrombin. There are many reports that dicumarol also inhibits NADPH:quinone oxidoreductase (NQO(1)). In one, it inhibited NQO(1) in a pancreatic cancer cell line, causing increased formation of superoxide and inhibiting cell growth.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Ann Nolan et al.
Biochemical pharmacology, 80(7), 977-981 (2010-07-06)
NAD(P)H quinone oxidoreductase (NQO1) has multiple functions in the cell including an ability to act as a detoxifying enzyme and as a protein chaperone. The latter property is particularly important in oncology as one of the client proteins of NQO1
Katherine Ann Scott et al.
Biochemical pharmacology, 81(3), 355-363 (2010-10-26)
The enzyme NAD(P)H quinone oxidoreductase (NQO1) can function both as a detoxifying enzyme as well as chaperone protein. The latter property has been extensively characterized by the use of dicoumarol which inhibits the chaperone properties of NQO1 in cells. However
Alexandra Reichstein et al.
Journal of medicinal chemistry, 55(16), 7273-7284 (2012-08-01)
A series of linearly anellated lapacho quinone analogues substituted at the 2-position of the tricyclic naphtho[2,3-b]furan-4,9-dione system were synthesized and evaluated for their ability to suppress keratinocyte hyperproliferation using HaCaT cells as the primary test system. While very good in
C O Lemley et al.
Journal of dairy science, 93(10), 4613-4624 (2010-09-22)
Progesterone is required for maintenance of pregnancy, and peripheral concentrations of progesterone are affected by both production and inactivation. Hepatic cytochrome P450 (EC 1.14.14.1) and aldo-keto reductase (EC 1.1.1.145-151) enzymes play a pivotal role in the first step of steroid
Ying Dong et al.
Cancer research, 70(20), 8088-8096 (2010-10-14)
The clinical experimental agent, β-lapachone (β-lap; Arq 501), can act as a potent radiosensitizer in vitro through an unknown mechanism. In this study, we analyzed the mechanism to determine whether β-lap may warrant clinical evaluation as a radiosensitizer. β-Lap killed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service