N8141
4-Nitroquinoline N-oxide
≥98%
Synonym(s):
4-NQO, 4-Nitroquinoline 1-oxide
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
C9H6N2O3
CAS Number:
Molecular Weight:
190.16
Beilstein:
165756
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98%
color
yellow to brown
mp
154-156 °C (lit.)
solubility
acetone: soluble, clear to hazy
storage temp.
−20°C
SMILES string
[O-][N+](=O)c1cc[n+]([O-])c2ccccc12
InChI
1S/C9H6N2O3/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H
InChI key
YHQDZJICGQWFHK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
4-Nitroquinoline N-oxide is a quinoline derivative widely used in chemical research and development.
Application
4-Nitroquinoline N-oxide has been used as a model compound to study its carcinogenic effects.
Biochem/physiol Actions
Skin and lung tumor initiator under experimental conditions.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Antitumor activity of ficus deltoidea extract on oral cancer: An in vivo study
Al-Koshab M, et al.
Journal of Oncology, 2020 (2020)
Interplay of DNA repair, homologous recombination, and DNA polymerases in resistance to the DNA damaging agent 4-nitroquinoline-1-oxide in Escherichia coli.
Williams AB, et al.
DNA Repair, 9(10), 1090-1097 (2010)
Ye Du et al.
Environment international, 135, 105369-105369 (2019-12-17)
Effluents from wastewater treatment plants (WWTPs) may contain various pollutants with potential toxic effects. Ozonation is widely applied to purify wastewater, which may influence the toxicity and water quality indices simultaneously. The main goal of this study was to reveal
The antimutagenic effect of monoterpenes against UV-irradiation-, 4NQO-and t-BOOH-induced mutagenesis in coli.
Nikolic B, et al.
Archives of Biological Sciences, 63(1), 117-128 (2011)
J Peter Svensson et al.
PloS one, 7(5), e37368-e37368 (2012-05-23)
Studies in Saccharomyces cerevisiae show that many proteins influence cellular survival upon exposure to DNA damaging agents. We hypothesized that human orthologs of these S. cerevisiae proteins would also be required for cellular survival after treatment with DNA damaging agents.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service